Skip to Content
Merck
All Photos(3)

Documents

D9154

Sigma-Aldrich

o-Dianisidine dihydrochloride

tablet

Synonym(s):

3,3′-Dimethoxybenzidine dihydrochloride, Fast Blue B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O2 · 2HCl
CAS Number:
Molecular Weight:
317.21
Beilstein:
3917996
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

o-Dianisidine dihydrochloride, tablet, 10 mg substrate per tablet

Quality Level

form

tablet

solubility

water: 1 tablet/10 mL, clear, colorless

storage temp.

2-8°C

SMILES string

Cl.Cl.COc1cc(ccc1N)-c2ccc(N)c(OC)c2

InChI

1S/C14H16N2O2.2ClH/c1-17-13-7-9(3-5-11(13)15)10-4-6-12(16)14(8-10)18-2;;/h3-8H,15-16H2,1-2H3;2*1H

InChI key

UXTIAFYTYOEQHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Specificity

o-Dianisidine (3,3′-dimethoxybenzidine) is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is yellow-orange in color and can be read spectrophotometrically at 405 nm. The reaction may be stopped with 5 M HCl.

Warning

Probably carcinogenic.

Reconstitution

Dissolve one tablet in 60 ml of 50 mM phosphate-citrate buffer, pH 5.0. Add 12 μl of fresh 30% hydrogen peroxide immediately before to use.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kaszuba et al.
Biochimica et biophysica acta, 1419(2), 221-228 (1999-07-17)
Reactive cationic and anionic liposomes have been prepared from mixtures of dimyristoylphosphatidylcholine (DMPC) and cholesterol incorporating dimethyldioctadecylammonium bromide and DMPC incorporating phosphatidylinositol, respectively. The liposomes were prepared by the vesicle extrusion technique and had the enzymes glucose oxidase (GO) encapsulated
Frédéric Frottin et al.
Molecular & cellular proteomics : MCP, 5(12), 2336-2349 (2006-09-12)
Methionine aminopeptidase (MAP) is a ubiquitous, essential enzyme involved in protein N-terminal methionine excision. According to the generally accepted cleavage rules for MAP, this enzyme cleaves all proteins with small side chains on the residue in the second position (P1')
Rémy Ricoux et al.
Bioconjugate chemistry, 19(4), 899-910 (2008-03-08)
To develop artificial hemoproteins that could lead to new selective oxidation biocatalysts, a strategy based on the insertion of various iron-porphyrin cofactors into Xylanase A (Xln10A) was chosen. This protein has a globally positive charge and a wide enough active
Cathrine Nordgaard et al.
International journal of molecular sciences, 22(17) (2021-09-11)
p38 and c-Jun N-terninal kinase (JNK) are activated in response to acute stress and inflammatory signals. Through modification of a plethora of substrates, these kinases profoundly re-shape cellular physiology for the optimal response to a harmful environment and/or an inflammatory
Wei Shen et al.
Biosensors & bioelectronics, 44, 171-176 (2013-02-22)
Herein we report a label-free microRNA (miRNA) biosensor in which the formation of a thin insulating film is used to amplify the analytical signal. Briefly, the biosensor is made of an oligonucleotide-coated gold electrode. After hybridizing with a target miRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service