Skip to Content
Merck
All Photos(1)

Key Documents

G3293

Supelco

Glucose Assay Reagent

for use with enzymatic assay kits GAHK20, SCA20, FA20, SA20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12164500
NACRES:
NA.84

analyte chemical class(es)

sugars (glucose)

technique(s)

photometry: suitable

impurities

≤0.005% Creatine phosphokinase, glutathione reductase, myokinase, NADH-oxidase,NADPH oxidase, phosphogluconic dehydrogenase, phosphoglucose isomerase, lactic dehydrogenase
<0.01% ATP-ase, myokinase

application(s)

food and beverages
general analytical

storage temp.

2-8°C

Specificity

< 0.05 % Activity resulting from cross-reactivity with the following substrates: fructose, dextran, sedoheptulose, arabinose, galactose, xylose, trehalose, mannose, sorbitol, +/- fucose or ribose.

Application

For the quantitative determination of glucose in food and other materials utilizing the hexokinase-glucose 6-phosphate dehydrogenase enzymatic assay.
Reconstitute with deionized water using volume indicated by package size (20 or 50 mL).

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michele Bertazzini et al.
Frontiers in plant science, 7, 288-288 (2016-03-26)
Numerous angiosperms rely on pollinators to ensure efficient flower fertilization, offering a reward consisting of nourishing nectars produced by specialized floral cells, known as nectaries. Nectar components are believed to derive from phloem sap that is enzymatically processed and transformed
Daniel S Straus
Molecular cancer, 12, 78-78 (2013-07-20)
Inflammation is a well-known etiological factor for colorectal cancer, but mechanisms underlying the linkage between inflammation and cancer are incompletely understood. We hypothesized that two pro-inflammatory cytokines, TNFα and IL-17, might play a role in promoting colorectal carcinogenesis. Aerobic glycolysis
Chao Zhang et al.
The Journal of animal ecology, 85(6), 1431-1441 (2016-10-26)
A key challenge for ecologists is to predict how single and joint effects of global warming and predation risk translate from the individual level up to ecosystem functions. Recently, stoichiometric theory linked these levels through changes in body stoichiometry, predicting
Mor Tsamir-Rimon et al.
The New phytologist, 229(3), 1398-1414 (2020-09-04)
Carbon reserve use is a major drought response in trees, enabling tree survival in conditions prohibiting photosynthesis. However, regulation of starch metabolism under drought at the whole-tree scale is still poorly understood. To this end, we combined measurements of nonstructural
Lara Kern et al.
Cell and tissue research, 366(3), 651-665 (2016-08-25)
Autophagy is a lysosomal degradation process involved in the turnover of organelles or other cell constituents, in providing sources for energy production under starving conditions and in cell metabolism. A key protein in the macroautophagic machinery is the autophagy-related protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service