Skip to Content
Merck
All Photos(2)

Documents

D1387

Sigma-Aldrich

P1,P3-Di(adenosine-5′) triphosphate ammonium salt

Synonym(s):

A(5′)P3(5′)A, Diadenosine triphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H24N9O16P3 · NH3
CAS Number:
Molecular Weight:
756.41
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51

form

solid

Quality Level

impurities

Other adenosine phosphates, essentially free

storage temp.

−20°C

SMILES string

N.Nc1ncnc2n(cnc12)C3OC(COP(O)(=O)OP(O)(=O)OP(O)(=O)OCC4OC(C(O)C4O)n5cnc6c(N)ncnc56)C(O)C3O

InChI

1S/C20H27N10O16P3.H3N/c21-15-9-17(25-3-23-15)29(5-27-9)19-13(33)11(31)7(43-19)1-41-47(35,36)45-49(39,40)46-48(37,38)42-2-8-12(32)14(34)20(44-8)30-6-28-10-16(22)24-4-26-18(10)30;/h3-8,11-14,19-20,31-34H,1-2H2,(H,35,36)(H,37,38)(H,39,40)(H2,21,23,25)(H2,22,24,26);1H3

InChI key

PKSQTRWRSNNYCW-UHFFFAOYSA-N

Related Categories

Application

P1,P3-Di(adenosine-5′) triphosphate (Ap3A) is a diadenosine oligophosphate (ApnA) family member. ApnAs have been found to act as a signaling molecule.
P1,P3-Di(adenosine-5′) triphosphate ammonium salt has been used as a component of the incubation medium for incubating fibroblast cell lines for FoF1 ATP-synthase activity.

Quantity

Approx. 25 A260 units per mg.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brigitte Maria Stavrou
Current medicinal chemistry. Cardiovascular and hematological agents, 1(2), 151-169 (2004-08-24)
Diadenosine polyphosphates (Ap(n)A) are naturally occurring molecules which have been identified in human and guinea-pig cardiac tissue and whose physiological role is being established. They are stored in platelet dense granules in a metabolically inactive form. Ap(n)A form a class
Michael Wright et al.
The FEBS journal, 273(15), 3534-3544 (2006-08-04)
Heat shock inducible lysyl-tRNA synthetase of Escherichia coli (LysU) is known to be a highly efficient diadenosine 5',5'''-P1,P4-tetraphosphate (Ap4A) synthase. However, we use an ion-exchange HPLC technique to demonstrate that active LysU mixtures actually have a dual catalytic activity, initially
Alan R Conant et al.
Vascular pharmacology, 48(4-6), 157-164 (2008-03-08)
Diadenosine polyphosphates (Ap(n)A) are released by degranulating platelets and high, local concentrations may form at sites of platelet activation. Radial artery grafts, now often used alongside the internal mammary artery in coronary artery bypass surgery, are particularly reactive to several
Dual roles of diadenosine polyphosphates in corneal epithelial cell migration
Mediero A, Peral A, Pintor J.
Investigative Ophthalmology & Visual Science, 47, 4500-4506 (2010)
Manuela Campiglio et al.
Journal of cellular physiology, 208(2), 274-281 (2006-03-21)
FHIT is a tumor suppressor gene that is frequently inactivated in human cancer. Although the Fhit protein is known to hydrolyze diadenosine triphosphate (Ap(3)A), this hydrolase activity is not required for Fhit-mediated oncosuppression. Indeed, the molecular mechanisms and the regulatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service