Skip to Content
Merck
All Photos(1)

Key Documents

01905

Sigma-Aldrich

Adenosine 5′-diphosphate

≥95% (HPLC)

Synonym(s):

5′-ADP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N5O10P2
CAS Number:
Molecular Weight:
427.20
Beilstein:
67722
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.55

Quality Level

Assay

≥95% (HPLC)

form

powder

impurities

≤15.0% water (KFT)

shipped in

dry ice

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O
Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O

InChI

1S/C10H15N5O10P2/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(24-10)1-23-27(21,22)25-26(18,19)20/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H2,11,12,13)(H2,18,19,20)/t4-,6-,7-,10-/m1/s1

InChI key

XTWYTFMLZFPYCI-KQYNXXCUSA-N

Gene Information

Application

Adenosine 5′-diphosphate (5′-ADP or ADP) was used as a test compound for studying the endothelium-dependent vascular response in salt sensitive (DS) and salt resistant Dahl rats (DR). The product was used to study the different P2-purinergic receptor subtypes on canine vascular smooth muscle and endothelium.

Biochem/physiol Actions

Adenosine 5′-diphosphate induces human platelet aggregation and non-competitively blocks the stimulated human platelet adenylate cyclase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T F Lüscher et al.
Hypertension (Dallas, Tex. : 1979), 9(2), 157-163 (1987-02-01)
Experiments were designed to study endothelium-dependent responses in salt-sensitive (DS) and salt-resistant Dahl rats (DR). The rats were fed a low sodium (0.1% NaCl) or high sodium (8% NaCl) diet for 8 weeks. Blood pressure in DS fed a high
R N Goldberg et al.
Biophysical chemistry, 40(3), 241-261 (1991-07-01)
The thermodynamic treatment of the disproportionation reaction of adenosine 5'-diphosphate to adenosine 5'-triphosphate and adenosine 5'-monophosphate is discussed in terms of an equilibrium model which includes the effects of the multiplicity of ionic and metal bound species and the presence
W G Mayhan
Inflammation, 16(4), 295-305 (1992-08-01)
The goal of this study was to determine the role of the synthesis and release of nitric oxide in modulating alterations in microvascular permeability of the hamster cheek pouch in response to adenosine 5'-diphosphate and bradykinin. We used intra-vital fluorescent
Yoshihiro Matsumura et al.
The Journal of biological chemistry, 288(43), 31069-31079 (2013-08-31)
The C terminus of Hsp70 interacting protein (CHIP) E3 ligase functions as a key regulator of protein quality control by binding the C-terminal (M/I)EEVD peptide motif of Hsp/c70(90) with its N-terminal tetratricopeptide repeat (TPR) domain and facilitating polyubiquitination of misfolded
N J Cusack et al.
British journal of pharmacology, 76(1), 221-227 (1982-05-01)
1 Adenosine 5'-diphosphate (ADP) induces human platelet aggregation and noncompetitively inhibits stimulated human platelet adenylate cyclase; it has been suggested that these two effects are mediated by separate ADP receptors on the platelet surface. 2 Adenosine 5'-triphosphate and seven adenine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service