CRISPR19
Lenti CRISPR Universal Non-Target Control#2 Plasmid DNA (LV04 vector)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106610
NACRES:
NA.51
Recommended Products
form
liquid
Quality Level
packaging
vial of 50 μL
concentration
20 ng/μL in TE buffer; DNA (1μg of plasmid DNA )
application(s)
CRISPR
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
The Lenti CRISPR Non-Target Control#2 (Lv04 vector) is a lentiviral plasmid vector, which includes a gRNA sequence that does not target known human, mouse and rat genes. The DNA format of this control construct permits direct transfection in knockout experiments or use in the viral packaging process (for lentiviral-based knockdown experiments). This vector is useful as a negative control in experiments using CRISPR lentiviral clones. The lenti CRISPR non-target control uses a dual-component system consisting of U6-driven guide RNA (non-targeting) and hPGK-driven Puromycin resistance cassette and Blue Fluorescence Protein for fluorophorescence. This control requires Cas9 be delivered by a separate vector.
Application
Functional Genomics/Screening/Target Validation
Other Notes
Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells, respectively. Self-inactivating replication incompetent viral particles can be produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids (Product No. SHP002).
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service