Skip to Content
Merck
All Photos(1)

Key Documents

SHP002

Sigma-Aldrich

CRISPR & MISSION® Lentiviral Packaging Mix

Synonym(s):

Lentiviral Packaging Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

concentration

100 ng/μL (in Tris-EDTA)

Quality Level

application(s)

CRISPR

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

The CRISPR & MISSION Lentiviral Packaging Mix is an optimized formulation of two plasmids expressing the key HIV packaging genes and a heterologous viral envelope gene.

Lentiviral particles are generated from three main components:
  • The packaging vector, which contains the minimal set of lentiviral genes required to generate the virion structural proteins and packaging functions.
  • The vesicular stomatitis virus G-protein envelope vector, which provides the heterologous envelope for pseudotyping and allows these lentiviral particles to efficiently deliver the transfer sequence of interest to a wide variety of cell types, including primary and non-dividing cells
  • The lentiviral transfer vector, which contains the sequence of interest as well as the cis acting sequences necessary for packaging.

The Lentiviral Packaging Mix contains the first two components; it is designed to be co-transfected along with a compatible lentiviral transfer vector in order to create high-titer pseudo-typed lentiviral particles used for downstream transduction applications. This product is recommended for packaging Lenti CRISPR, Lenti ORF, and Lenti shRNA vectors.

Application

Functional Genomics/Screening/Target Validation

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service