Skip to Content
Merck
All Photos(1)

Key Documents

CRISPR12

Sigma-Aldrich

CRISPR-Lenti Non-Targeting Control Plasmid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

form

liquid

Quality Level

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

The Sigma lenti CRISPR Non-Targeting Control is a lentiviral plasmid vector, which includes a gRNA sequence that does not target known human, mouse and rat genes. This vector is useful as a negative control in experiments using Sigma CRISPR lentiviral clones. The Sigma lenti CRISPR non-targeting control uses a one-plasmid system consisting of a EF1-alpha-driven Cas9, a U6-driven guide RNA (non-targeting), with both Puromycin and GFP co-expressed with Cas9.

Ampicillin and puromycin antibiotic resistance genes provide selection in bacterial or mammalian cells, respectively. Also, self-inactivating replication incompetent viral particles can be produced in packaging cells (HEK293T) by co-transfection with compatible packaging plasmids (Product No. SHP001).

Clustered regularly interspaced short palindromic repeats (CRISPR)/CRISPR-associated (Cas), originally found in bacteria and archaea, is employed by organisms to protect them from invading viruses and plasmids.

Application

Functional Genomics/Screening/Target Validation
  • Non-targeting control, CRISPR lentivirus DNA format, for creating gene knockouts/genetic modifications in cell lines.

To see more application data, protocols, vector maps visit sigma.com/crisprs.

Features and Benefits

Serves as a non-targeting or negative experimental control (DNA format) for the CRISPR editing workflow using lenti CRISPRs. This allows you to examine the effect of transfection in your system with the Sigma lenti CRISPR/Cas9 plasmid.

Physical form

The Sigma CRISPR Lenti Non-Targeting Control Vector is provided as 1 μg of plasmid DNA in Tris-EDTA (TE).

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Basit Salik et al.
Cancer cell, 38(2), 263-278 (2020-06-20)
Signals driving aberrant self-renewal in the heterogeneous leukemia stem cell (LSC) pool determine aggressiveness of acute myeloid leukemia (AML). We report that a positive modulator of canonical WNT signaling pathway, RSPO-LGR4, upregulates key self-renewal genes and is essential for LSC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service