Skip to Content
Merck
All Photos(1)

Documents

C1655

Sigma-Aldrich

Cholera Toxin B subunit

FITC conjugate, lyophilized powder

Synonym(s):

CTB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

conjugate

FITC conjugate

Quality Level

form

lyophilized powder

mol wt

~12 kDa

composition

Protein, ~20% Lowry

extent of labeling

~1.0 mol FITC per mol protein

storage temp.

2-8°C

SMILES string

CCOc1ccccc1C(=O)Nc2ccc(Cl)c(c2)C(F)(F)F

InChI

1S/C16H13ClF3NO2/c1-2-23-14-6-4-3-5-11(14)15(22)21-10-7-8-13(17)12(9-10)16(18,19)20/h3-9H,2H2,1H3,(H,21,22)

InChI key

YDXZSNHARVUYNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cholera Toxin B subunit-fluorescein isothiocyanate (FITC) conjugate has been used as a fluorescent tracer:
  • to label distal cerebrospinal fluid-contacting neurons (dCSF-CNs) in lateral ventricles of rat
  • to stain epididymal sperms of mice
  • to stain peritoneal macrophages of mice

Biochem/physiol Actions

The cholera toxin B subunit is used for track tracing in neurological research, taking advantage of GM1 ganglioside binding and retrograde transport. Tissue culture cells treated with cholera toxin are not killed and tissues of animals do not become necrotic.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Physical form

Lyophilized powder containing Tris buffer salts, sodium chloride, sodium EDTA and sodium azide

Analysis Note

Activity measured by ELISA using ganglioside GM1 coated multiwell plates, rabbit anti-cholera toxin B subunit antibodies, and peroxidase-labeled goat anti-rabbit IgG as the second antibody. 50% saturation of binding was achieved with 0.01-1 μg of the cholera toxin B subunit-FITC conjugate per mL.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily Igel et al.
The Journal of biological chemistry, 297(3), 101106-101106 (2021-08-24)
Polymorphisms in the apolipoprotein E (apoE) gene are risk factors for chronic inflammatory diseases including atherosclerosis. The gene product apoE is synthesized in many cell types and has both lipid transport-dependent and lipid transport-independent functions. Previous studies have shown that
Lindsey C Felix et al.
Aquatic toxicology (Amsterdam, Netherlands), 192, 58-68 (2017-09-18)
The ever-growing production of engineered nanoparticles (NPs) for use in many agricultural, commercial, consumer, and industrial applications will lead to their accidental or intentional release into the environment. Potential routes of environmental exposure include manufacturing or transport spills, disposal of
Laura Aragoneses-Fenoll et al.
Journal of leukocyte biology (2021-02-03)
The interaction between the T-lymphocyte costimulatory molecule ICOS and its ligand (ICOS-L) is needed for efficient immune responses, but expression levels are tightly controlled, as altered expression of ICOS or ICOS-L may lead to immunodeficiency, or favor autoimmune diseases and
Jill M Steinbach et al.
Acta biomaterialia, 30, 49-61 (2015-11-26)
The surface modification of nanoparticles (NPs) can enhance the intracellular delivery of drugs, proteins, and genetic agents. Here we studied the effect of different surface ligands, including cell penetrating peptides (CPPs), on the cell binding and internalization of poly(lactic-co-glycolic) (PLGA)
Min Huang et al.
Oncology letters, 21(6), 438-438 (2021-04-20)
Pancreatic cancer is a highly malignant type of cancer and its treatment remains a major challenge. The novel recombinant protein TNF-related apoptosis-inducing ligand (TRAIL)-Mu3 has been shown to exert stronger tumor inhibitory effects in colon cancer in vitro and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service