Skip to Content
Merck
All Photos(1)

Key Documents

C7344

Sigma-Aldrich

Cyclic adenosine diphosphate-ribose

≥90% (HPLC), lyophilized powder

Synonym(s):

ADP-cyclo[N1:1′′]-ribose, Cyclic ADPR, cADP-ribose, cADPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N5O13P2
CAS Number:
Molecular Weight:
541.30
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51

biological source

(Ovotestis from sea hare)

Quality Level

Assay

≥90% (HPLC)

form

lyophilized powder

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](O)[C@H]2O[C@@H]1COP(O)(=O)OP(O)(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)[n@H]4cnc5C(=N)[N@@H]2C=Nc45

InChI

1S/C15H21N5O13P2/c16-12-7-13-18-4-19(12)14-10(23)8(21)5(31-14)1-29-34(25,26)33-35(27,28)30-2-6-9(22)11(24)15(32-6)20(13)3-17-7/h3-6,8-11,14-16,21-24H,1-2H2,(H,25,26)(H,27,28)/t5-,6-,8-,9-,10-,11-,14-,15-/m1/s1

InChI key

BQOHYSXSASDCEA-KEOHHSTQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyclic adenosine diphosphate-ribose (cADPR) is an intracellular second messenger that mobilizes intracellular Ca2+ release from the sarcoplasmic reticulum through ryanodine receptors. CADPR is also a second messenger for glucose-induced insulin release from endocrine pancreas and for abscisic acid-induced calcium release in plants.
Cyclic adenosine diphosphate-ribose has been used to stimulate astrocytes to activate CD38 signaling. It has also been used as a supplement in buffered peptone water to recover S. enteritidis.

Biochem/physiol Actions

Mobilizes intracellular Ca2+; regulates Ca2+-induced Ca2+ release specifically blocked by ryanodine; second messenger for glucose-induced insulin release from the endocrine pancreas and for abscisic acid-induced calcium release in plants.

Packaging

Package size based on nucleotide content.

Physical form

Lyophilized powder

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hon Cheung Lee
Science China. Life sciences, 54(8), 699-711 (2011-07-26)
The concept advanced by Berridge and colleagues that intracellular Ca(2+)-stores can be mobilized in an agonist-dependent and messenger (IP(3))-mediated manner has put Ca(2+)-mobilization at the center stage of signal transduction mechanisms. During the late 1980s, we showed that Ca(2+)-stores can
Shunichi Kajioka et al.
The Journal of urology, 186(2), 736-744 (2011-06-21)
To clarify the properties of adenosine triphosphate sensitive K+ channel in human detrusor smooth muscle we examined the effect of the representative nicotinic acid derivatives β-nicotinamide adenine dinucleotide, cyclic adenosine diphosphate ribose and nicotinic acid adenine dinucleotide phosphate (Sigma-Aldrich®) on
Y Wu et al.
Science (New York, N.Y.), 278(5346), 2126-2130 (1998-02-12)
Abscisic acid (ABA) is the primary hormone that mediates plant responses to stresses such as cold, drought, and salinity. Single-cell microinjection experiments in tomato were used to identify possible intermediates involved in ABA signal transduction. Cyclic ADP-ribose (cADPR) was identified
Transfer of mitochondria from astrocytes to neurons after stroke
Hayakawa K, et al.
Nature, 535(7613), 551-551 (2016)
Jing Xiong et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 31(1), 44-55 (2013-01-25)
Despite extensive studies, the intracellular regulatory mechanism of renin production and release is still poorly understood. The present study was designed to test whether CD38-ADP-ribosylcyclase signaling pathway contributes to the regulation of renin production and release, and to examine whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service