Skip to Content
Merck
All Photos(2)

Key Documents

N7764

Sigma-Aldrich

Nicotinic acid mononucleotide

Synonym(s):

Nicotinate mononucleotide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14NO9P
CAS Number:
Molecular Weight:
335.20
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.79

biological source

animal

Assay

≥98.0% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white

storage temp.

−20°C

SMILES string

OC1C(O)C(OC1COP(O)(O)=O)[N]2=CC(=CC=C2)C(O)=O

InChI

1S/C11H15NO9P/c13-8-7(5-20-22(17,18)19)21-10(9(8)14)12-3-1-2-6(4-12)11(15)16/h1-4,7-10,13-14H,5H2,(H,15,16)(H2,17,18,19)

InChI key

IUJWGLOJJKFASX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinic acid mononucleotide (NaMN) is formed from nicotinic acid 5-phosphoribosyl-1-pyrophosphate (PRPP) in the Preiss-Handler pathway in the presence of enzyme phosphoribosyltransferase (NaPRT).

Application

Nicotinic acid mononucleotide has been used as a substrate for NMN/NaMN adenylyltransferase (NMNAT) during nicotinamide adenine dinucleotide (NAD) biosynthesis.

Biochem/physiol Actions

Nicotinic acid mononucleotide (NaMN) may be used to study the relationship between the phosphate-responsive signaling (PHO) pathway and nicotine adenine dinucleotide (NAD(+)) metabolism in yeast. Nicotinate mononucleotide is a substrate for NMN/NaMN adenylyltransferase (NMNAT) and nicotinate mononucleotide adenylyltransferase. NaMN amidation results in the synthesis of nicotinamide mononucleotide(NMN), which is further converted to NAD cofactor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Sun et al.
Journal of bacteriology, 175(5), 1423-1432 (1993-03-01)
A number of Bacillus subtilis genes involved in NAD biosynthesis have been cloned and sequenced. One of the genes encodes a polypeptide homologous to Escherichia coli L-aspartate oxidase, and its mutation resulted in a nicotinic acid (Nic)-dependent phenotype; this gene
William B Black et al.
Microbial cell factories, 19(1), 150-150 (2020-07-29)
Noncanonical redox cofactors are emerging as important tools in cell-free biosynthesis to increase the economic viability, to enable exquisite control, and to expand the range of chemistries accessible. However, these noncanonical redox cofactors need to be biologically synthesized to achieve
Kazuo Yamada et al.
Analytical biochemistry, 352(2), 282-285 (2006-04-01)
We have developed a liquid chromatographic-tandem mass spectrometric method that is sensitive and specific and that simultaneously measures cellular NAD(+) and related compounds. Using this method, NAD(+), NAAD, NMN, NAMN, NAM, NA, ADPR, and 5'AMP were first separated over a
Determining NAD synthesis in erythrocytes.
V Micheli et al.
Methods in enzymology, 280, 211-221 (1997-01-01)
Stimulation of nicotinamide adenine dinucleotide biosynthetic pathways delays axonal degeneration after axotomy
Sasaki Y, et al.
Journal of neurosciences in rural practice, 26(33), 8484-8491 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service