Skip to Content
Merck
All Photos(1)

Key Documents

C6240

Sigma-Aldrich

CITCO

≥98% (HPLC), solid

Synonym(s):

6-(4-Chlorophenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde O-(3,4-dichlorobenzyl)oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12N3OCl3S
CAS Number:
Molecular Weight:
436.74
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

DMSO: soluble 28 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

Clc1ccc(cc1)-c2nc3sccn3c2\C=N\OCc4ccc(Cl)c(Cl)c4

InChI

1S/C19H12Cl3N3OS/c20-14-4-2-13(3-5-14)18-17(25-7-8-27-19(25)24-18)10-23-26-11-12-1-6-15(21)16(22)9-12/h1-10H,11H2/b23-10+

InChI key

ZQWBOKJVVYNKTL-AUEPDCJTSA-N

General description

CITCO is an imidazothiazole derivative. It stimulates human constitutive androstane receptor (CAR) nuclear translocation.

Application

CITCO has been used for the activation of mouse constitutive androstane receptor (CAR) and human CAR.

Biochem/physiol Actions

CITCO is a constitutive androstane receptor (CAR) agonist; nuclear receptor NR113 agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a novel human constitutive androstane receptor (CAR) agonist and its use in the identification of CAR target genes
Maglich JM, et al.
The Journal of Biological Chemistry, 278(19), 17277-17283 (2003)
Won-Gu Choi et al.
Pharmaceutics, 12(3) (2020-03-07)
Mertansine, a tubulin inhibitor, is used as the cytotoxic component of antibody-drug conjugates (ADCs) for cancer therapy. The effects of mertansine on uridine 5'-diphospho-glucuronosyltransferase (UGT) activities in human liver microsomes and its effects on the mRNA expression of cytochrome P450s
Constitutive androstane receptor (Car)-driven regeneration protects liver from failure following tissue loss
Tschuor C, et l.
Journal of Hepatology, 65(1), 66-74 (2016)
Tadeja Režen et al.
Drug metabolism and disposition: the biological fate of chemicals, 45(8), 974-976 (2017-05-26)
Statins are well known lipid lowering agents that inhibit the enzyme 3-hydroxy-3-methylglutaryl-CoA (HMG-CoA) reductase. They also activate drug metabolism but their exact receptor-mediated action has not been proven so far. We tested whether atorvastatin and rosuvastatin are direct ligands of
Constitutive androstane receptor agonist CITCO inhibits growth and expansion of brain tumour stem cells
Chakraborty S , et al.
British Journal of Cancer, 104(3), 448-448 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service