Skip to Content
Merck
All Photos(3)

Key Documents

P1636

Sigma-Aldrich

Phenobarbital

Synonym(s):

5-Ethyl-5-phenyl-2,4,6-pyrimidinetrione, 5-Ethyl-5-phenylbarbituric acid, Luminal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O3
CAS Number:
Molecular Weight:
232.24
Beilstein:
233363
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

drug control

USDEA Schedule IV; Home Office Schedule 3; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

originator

Bayer

SMILES string

CCC1(C(=O)NC(=O)NC1=O)c2ccccc2

InChI

1S/C12H12N2O3/c1-2-12(8-6-4-3-5-7-8)9(15)13-11(17)14-10(12)16/h3-7H,2H2,1H3,(H2,13,14,15,16,17)

InChI key

DDBREPKUVSBGFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anesthetic; sedative; hypnotic; anticonvulsant

Features and Benefits

This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Noura M Darwish et al.
Molecules (Basel, Switzerland), 27(9) (2022-05-15)
Hepatocellular carcinoma (HCC) is a common type of liver cancer and is a leading cause of death worldwide. Signal transducer and activator of transcription 3 (STAT3) is involved in HCC progression, migration, and suppression of apoptosis. This study investigates the
Peter J West et al.
Experimental neurology, 349, 113954-113954 (2021-12-20)
The discovery and development of novel antiseizure drugs (ASDs) that are effective in controlling pharmacoresistant spontaneous recurrent seizures (SRSs) continues to represent a significant unmet clinical need. The Epilepsy Therapy Screening Program (ETSP) has undertaken efforts to address this need
Melissa Barker-Haliski et al.
Epilepsia, 62(7), 1677-1688 (2021-06-04)
The kainic acid (KA)-induced status epilepticus (SE) model in rats is a well-defined model of epileptogenesis. This model closely recapitulates many of the clinical and pathological characteristics of human temporal lobe epilepsy (TLE) that arise following SE or another neurological
M Lundqvist et al.
Acta paediatrica (Oslo, Norway : 1992), 102(9), 863-867 (2013-06-07)
Treatment of neonatal seizures still relies primarily on phenobarbital, despite an estimated efficacy of less than 50% and concern over neurodegenerative side effects. The objective of this study was to evaluate the efficacy and safety of lidocaine as second-line treatment
Martin J Brodie et al.
Epilepsia, 53 Suppl 8, 40-46 (2012-12-12)
This article reviews the current position of phenobarbital using articles published since 2000 and speculates on its likely future contribution to epilepsy care. Over the last decade there have been no major double-blind randomized placebo-controlled or comparative trials with phenobarbital.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service