Skip to Content
Merck

Skip To

AB152

Anti-Tyrosine Hydroxylase Antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

Tyrosine 3-hydroxylase, Tyrosine 3-monooxygenase, tyrosine hydroxylase, TH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
polyclonal
Species reactivity:
human, mollusk, feline, ferret, Aplysia (marine mollusc), rat, squid, Drosophila, mouse
Application:
ELISA, IF, IHC (p), IP, WB
Citations:
1287
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit

biological source

rabbit

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mollusk, feline, ferret, Aplysia (marine mollusc), rat, squid, Drosophila, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunofluorescence: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TH(7054)
rat ... Th(25085)

General description

62 kDa
Tyrosine hydroxylase (also know as Tyrosine 3-hydroxylase and Tyrosine 3-monooxygenase) plays an important role in the physiology of adrenergic neurons. It is the first and rate-limiting enzyme involved in the biosynthesis of the catecholamines Dopamine and Norepinephrine from tyrosine. TH is, therefore, a useful marker for dopaminergic and noradrenergic neurons. The enzymatic activity of TH requires ferrous ions as cofactors and is believed to be regulated by phosphorylation. At least four isoforms of human TH have been identified which result from alternative splicing.

Immunogen

Denatured tyrosine hydroxylase from rat pheochromocytoma (denatured by sodium dodecyl sulfate).

Application

Anti-Tyrosine Hydroxylase Antibody detects level of TH and has been published and validated for use in ELISA, IF, IH, IH(P), IP and WB.
Immunohistochemistry:
A previous lot of this antibody was uesd at 1:100-1:1,000 dilution, 4°C, for 1-3 days. Stains both fresh frozen and paraffin embedded tissue samples by indirect immunofluorescence and immunoperoxidase.

Immunofluorescence:
A previous lot of this antibody was used at 1:100-1:1,000 dilution.

Immunoprecipitation:
A previous lot of this antibody was used on Immunoprecipitation.

ELISA:
A previous lot of this antibody was used in ELISA.

Optimal dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers

Biochem/physiol Actions

Anti-Tyrosine Hydroxylase Antibodies react with most mammalian and many non-mammalian species. It has been reported that this antibody does not work on paraffin-embedded human tissue
Tyrosine hydroxylase. By western blot, AB152 selective labels a single band at approximately 62kDa (reduced) corresponding to Tyrosine Hydroxylase. It is expected that the antibody will react with most mammalian and many non-mammalian species. It has been reported that this antibody does not work on paraffin embedded human tissue.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing liquid in 10 mM HEPES (pH 7.5), 150 mM NaCl (pH 7.5), 100 ug/mL BSA and 50% glycerol.

Preparation Note

Maintain frozen at -20ºC to -70ºC in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles. Do not store in a self defrosting freezer.

Analysis Note

Control
POSITIVE CONTROL: Brain (corpus striatum, sympathetic nerve terminals) and adrenal glands.
NEGATATIVE CONTROL: Liver.
Routinely evaluated by Western Blot on PC12 lysates.

Western Blot Analysis:
1:1000 dilution of this lot detected tyrosine hydroxylase on 10 μg of PC12 lysates.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB5935MAB318SAB4200697
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

ascites fluid

antibody form

purified immunoglobulin

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

mouse

Gene Information

human ... TH(7054)
rat ... Th(25085)

Gene Information

human ... TH(7054)
rat ... Th(25085)

Gene Information

human ... TH(7054)
rat ... Th(25085)

Gene Information

human ... TH(7054)

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

clone

polyclonal

clone

polyclonal

clone

LNC1, monoclonal

clone

TH-16, monoclonal


Still not finding the right product?

Explore all of our products under Anti-Tyrosine Hydroxylase Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Hello! What is the concentration in mg/ml of AB152?

    1 answer
    1. The concentration of this product is 150 µg/mL.

      Helpful?

  2. Greetings, I would like to know if this Antibody can be used for immune cytochemistry procedure with dopaminergic cell line N27-A

    1 answer
    1. As it shows in the link below, MAB318-AF488 has been used for detection of tyrosine hydroxylase in SCC196. Theoretically AB152 should work but it has not been tested in house.
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/130/252/scc196ds.pdf

      Helpful?

Reviews

No rating value

Active Filters