Skip to Content
Merck
All Photos(5)

Key Documents

C3032

Sigma-Aldrich

Cesium chloride

for molecular biology, ≥99% (silver nitrate titration)

Synonym(s):

CsCl, cesium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.31
Assay:
≥99% (silver nitrate titration)
grade:
for molecular biology
form:
powder
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

Assay

≥99% (silver nitrate titration)

form

powder

impurities

≤0.001% Li
≤0.01% Pb

mp

645 °C (lit.)

absorption

≤0.02 at 280 nm at 50%
≤0.1 at 260 nm at 50%

foreign activity

DNase exonuclease, Nickase endonuclease, RNase, protease, none detected

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Used for the preparation of electrically conducting glasses.Used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuan A Ho et al.
Langmuir : the ACS journal of surfaces and colloids, 28(2), 1256-1266 (2011-12-14)
All-atom molecular dynamics simulations were conducted to study the dynamics of aqueous electrolyte solutions confined in slit-shaped silica nanopores of various degrees of protonation. Five degrees of protonation were prepared by randomly removing surface hydrogen atoms from fully protonated crystalline
Takafumi Yamamoto et al.
Inorganic chemistry, 50(22), 11787-11794 (2011-10-25)
The rock salt (B1) structure of binary oxides or chalcogenides transforms to the CsCl (B2) structure under high pressure, with critical pressures P(s) depending on the cation to anion size ratio (R(c)/R(a)). We investigated structural changes of A(2)MO(3) (A =
Piotr Jurkiewicz et al.
Biochimica et biophysica acta, 1818(3), 609-616 (2011-12-14)
Effects of alkali metal chlorides on the properties of mixed negatively charged lipid bilayers are experimentally measured and numerically simulated. Addition of 20mol% of negatively charged phosphatidylserine to zwitterionic phosphatidylcholine strengthens adsorption of monovalent cations revealing their specificity, in the
Travis A Hage et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(8), 2714-2721 (2012-02-24)
We report a novel coupled system of sodium-activated potassium currents (I(KNa)) and persistent sodium currents (I(NaP)), the components of which are widely distributed throughout the brain. Its existence and importance has not been previously recognized. Although I(KNa) was known to
Sandra Aedo et al.
Antimicrobial agents and chemotherapy, 56(11), 5458-5464 (2012-08-08)
DNA topoisomerases are important targets in anticancer and antibacterial therapy because drugs can initiate cell death by stabilizing the transient covalent topoisomerase-DNA complex. In this study, we employed a method that uses CsCl density gradient centrifugation to separate unbound from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service