Skip to Content
Merck
All Photos(3)

Key Documents

C2786

Sigma-Aldrich

Calcium nitrate tetrahydrate

suitable for cell culture, suitable for plant cell culture

Synonym(s):

Ca(NO3)2.4H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
236.15
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Quality Level

form

solid

technique(s)

cell culture | mammalian: suitable
cell culture | plant: suitable

mp

44 °C (dec.) (lit.)

SMILES string

O.O.O.O.[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ca.2NO3.4H2O/c;2*2-1(3)4;;;;/h;;;4*1H2/q+2;2*-1;;;;

InChI key

ICSSIKVYVJQJND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Production of Nano Hydroxyapatite and Mg-Whitlockite from Biowaste-Derived Products via Continuous Flow Hydrothermal Synthesis: A Step towards Circular Economy.: The research discusses the continuous flow hydrothermal synthesis of nano hydroxyapatite and Mg-whitlockite using calcium nitrate tetrahydrate. The process highlights the potential for sustainable production of bioceramics from biowaste (Nigar et al., 2023).

  • Preparation and In Vitro Bioactivity Study of a Novel Hollow Mesoporous Bioactive Glass Nanofiber Scaffold.: This paper details the preparation of a novel bioactive glass nanofiber scaffold using calcium nitrate tetrahydrate, highlighting its bioactivity and potential for tissue engineering applications (Xiao et al., 2022).

  • Bioinspired Nanonetwork Hydroxyapatite from Block Copolymer Templated Synthesis for Mechanical Metamaterials.: The research demonstrates the use of calcium nitrate tetrahydrate in the synthesis of bioinspired nanonetwork hydroxyapatite, emphasizing its mechanical properties and potential applications in advanced materials (Sadek et al., 2022).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehdi Razavi et al.
Materials science & engineering. C, Materials for biological applications, 48, 21-27 (2015-01-13)
Although magnesium (Mg) is a unique biodegradable metal which possesses mechanical property similar to that of the natural bone and can be an attractive material to be used as orthopedic implants, its quick corrosion rate restricts its actual clinical applications.
Da-Young Seong et al.
Journal of photochemistry and photobiology. B, Biology, 146, 34-43 (2015-03-22)
Although methylene blue (MB) is the most inexpensive photosensitizer with promising applications in the photodynamic therapy (PDT) for its high quantum yield of singlet oxygen generation, the clinical use of MB has been limited by its rapid enzymatic reduction in
Senthilguru Kulanthaivel et al.
Materials science & engineering. C, Materials for biological applications, 58, 648-658 (2015-10-20)
The present study delineates the synthesis and characterization of cobalt doped proangiogenic-osteogenic hydroxyapatite. Hydroxyapatite samples, doped with varying concentrations of bivalent cobalt (Co(2+)) were prepared by the ammoniacal precipitation method and the extent of doping was measured by ICP-OES. The
Kalimuthu Pandi et al.
Carbohydrate polymers, 134, 732-739 (2015-10-03)
Adsorption is a significant reaction occurs between adsorbent/water interface for controlling the pollutants in the aqueous environment. In this regard, an eco-magnetic biosorbent was prepared by uniform deposition of magnetic Fe3O4 particles on the surface of nano-hydroxyapatite (n-HAp)/chitosan (CS) nanocomposite
Cong Mao et al.
Biomedical materials (Bristol, England), 10(2), 025005-025005 (2015-03-26)
The ability of biomaterials to induce rapid vascular formation is critical in tissue regeneration. Combining recombinant angiogenic growth factors with bioengineered constructs have proven to be difficult due to several issues, including the instability of recombinant proteins, the need for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service