Skip to Content
Merck
All Photos(3)

Key Documents

C4955

Sigma-Aldrich

Calcium nitrate tetrahydrate

BioXtra, ≥99.0%

Synonym(s):

Ca(NO3)2, Calcium dinitrate, Calcium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
236.15
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
form:
powder
solubility:
H2O: 1 M at 20 °C, clear, colorless

grade

for analytical purposes

Quality Level

product line

BioXtra

Assay

≥99.0%

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

mp

44 °C (dec.) (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.002%

cation traces

Al: ≤0.002%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.01%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.O.O.O.[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ca.2NO3.4H2O/c;2*2-1(3)4;;;;/h;;;4*1H2/q+2;2*-1;;;;

InChI key

ICSSIKVYVJQJND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Exploration of Whitlockite Nanostructures for Hemostatic Applications.: This study explores the use of Whitlockite nanostructures, synthesized using calcium nitrate tetrahydrate, for their potential in hemostatic applications. The findings suggest that these nanostructures can significantly enhance blood clotting, making them a viable option for medical use (Jain et al., 2024).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehdi Razavi et al.
Materials science & engineering. C, Materials for biological applications, 48, 21-27 (2015-01-13)
Although magnesium (Mg) is a unique biodegradable metal which possesses mechanical property similar to that of the natural bone and can be an attractive material to be used as orthopedic implants, its quick corrosion rate restricts its actual clinical applications.
Miho Nakamura et al.
Biomedical materials (Bristol, England), 10(1), 011001-011001 (2015-01-15)
The surface modification of ceramic biomaterials used for medical devices is expected to improve osteoconductivity through control of the interfaces between the materials and living tissues. Polarisation treatment induced surface charges on hydroxyapatite, β-tricalcium phosphate, carbonate-substituted hydroxyapatite and yttria-stabilized zirconia
Véronique Lenoble et al.
Journal of colloid and interface science, 448, 473-481 (2015-03-17)
Three novel Ni(II)-Ion-Imprinted Polymer (IIP) were synthesized by precipitation polymerization of ethylene glycol dimethacrylate (crosslinker) with a complex of nickel(II) and vinylbenzyl iminodiacetic acid (VbIDA). The three IIPs were prepared with various mixtures of porogen solvents: methanol, methanol/2-methoxyethanol and methanol/acetonitrile
Da-Young Seong et al.
Journal of photochemistry and photobiology. B, Biology, 146, 34-43 (2015-03-22)
Although methylene blue (MB) is the most inexpensive photosensitizer with promising applications in the photodynamic therapy (PDT) for its high quantum yield of singlet oxygen generation, the clinical use of MB has been limited by its rapid enzymatic reduction in
Kalimuthu Pandi et al.
Carbohydrate polymers, 134, 732-739 (2015-10-03)
Adsorption is a significant reaction occurs between adsorbent/water interface for controlling the pollutants in the aqueous environment. In this regard, an eco-magnetic biosorbent was prepared by uniform deposition of magnetic Fe3O4 particles on the surface of nano-hydroxyapatite (n-HAp)/chitosan (CS) nanocomposite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service