Skip to Content
Merck
All Photos(1)

Key Documents

A3795

Sigma-Aldrich

Ammonium nitrate

suitable for plant cell culture

Synonym(s):

Ammonia nitrate, NH4NO3 , Nitric acid ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4NO3
CAS Number:
Molecular Weight:
80.04
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.71

Assay

≥99.5% (titration)

Quality Level

form

powder, crystals or chunks

technique(s)

cell culture | plant: suitable

bp

210 °C (lit.)

mp

169 °C (lit.)

SMILES string

N.O[N+]([O-])=O

InChI

1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3

InChI key

PRORZGWHZXZQMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Fertilizer Research: Ammonium nitrate is pivotal in agricultural studies, particularly in optimizing the nitrogen cycle and nutrient management in soil to enhance crop yields and minimize environmental impact. Its effectiveness as a nitrogen source in fertilizer formulations continues to be a key focus in agricultural sciences (Śniegowska et al., 2024).

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1C - Ammonium nitrate and ammonium nitrate containing preparations

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yung-Lin Hsieh et al.
Cell reports, 29(8), 2134-2143 (2019-11-21)
Amyotrophic lateral sclerosis (ALS), the most common motor neuron disease, usually occurs in middle-aged people. However, the molecular basis of age-related cumulative stress in ALS pathogenesis remains elusive. Here, we found that mice deficient in NPGPx (GPx7), an oxidative stress
Mohammad Mahdi Najafpour et al.
Dalton transactions (Cambridge, England : 2003), 41(34), 10156-10160 (2012-07-27)
Nanoscale manganese oxides within Faujasite zeolite have been synthesized with a simple method and characterized by scanning electron microscopy, X-ray diffraction spectrometry, N(2) adsorption-desorption isotherms, transmission electron microscopy, and atomic absorption spectroscopy. These oxides showed efficient water oxidizing activity in
Sari Palmroth et al.
Annals of botany, 111(3), 467-477 (2013-01-10)
Water and nitrogen (N) are two limiting resources for biomass production of terrestrial vegetation. Water losses in transpiration (E) can be decreased by reducing leaf stomatal conductance (g(s)) at the expense of lowering CO(2) uptake (A), resulting in increased water-use
Deborah Wakeham et al.
Physical chemistry chemical physics : PCCP, 14(15), 5106-5114 (2012-01-31)
The nanostructure of the ethanolammonium nitrate (EtAN)-air surface has been investigated using X-ray reflectometry (XRR), vibrational sum frequency spectroscopy (VSFS) and neutral impact collision ion scattering spectroscopy (NICISS). The XRR data decays more rapidly than expected for a perfectly sharp
Jason K Kirby et al.
Chemosphere, 89(7), 876-883 (2012-06-19)
Aging reactions in soils can influence the lability and hence bioavailability of added metals in soils through their removal from labile pools into pools from which desorption is slow (non-labile pools). The aims of this study were to examine the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service