Skip to Content
Merck
All Photos(2)

Key Documents

F8508

Sigma-Aldrich

Iron(III) nitrate nonahydrate

BioReagent, suitable for cell culture

Synonym(s):

Ferric nitrate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
404.00
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71

product line

BioReagent

Assay

98-101%

technique(s)

cell culture | mammalian: suitable

mp

47 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Fe.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SZQUEWJRBJDHSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iron(II) sulfate heptahydrate has useful cell culture applications. Thermal decomposition of iron(III) nitrate nonahydrate has been reported.

Application


  • Chemistry of iron nitrate-based precursor solutions for spray-flame synthesis: This study explores the chemical properties and solubility behavior of iron(III) nitrate nonahydrate in the context of producing nanoparticles via spray-flame synthesis, presenting a method that could advance material fabrication technologies (Stodt et al., 2019).

  • Development of Nitrolactonization Mediated by Iron(III) Nitrate Nonahydrate: This paper introduces a novel synthetic application of iron(III) nitrate nonahydrate in the nitrolactonization process, enhancing the methodology for constructing lactone frameworks in organic chemistry (Yoshimura et al., 2020).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silica supported ferric nitrate nonahydrate: selective oxidation of benzoins under mild conditions.
Paul AM, et al.
Synthetic Communications, 33(15), 2581-2584 (2003)
Thermal decomposition of Ni (II) and Fe (III) nitrates and their mixture.
Elmasry MAA, et al.
Journal of Thermal Analysis and Calorimetry, 52(2), 489-495 (1998)
Synthesis of iron (III)-doped titania nanoparticles and its application for photodegradation of sulforhodamine-B pollutant.
Kumbhar A and Chumanov G.
Journal of Nanoparticle Research, 7(4-5), 489-498 (2005)
Yu Jin Park et al.
Journal of translational medicine, 18(1), 236-236 (2020-06-14)
The in vitro production of mature human red blood cells (RBCs) from induced pluripotent stem cells (iPSCs) has been the focus of research to meet the high demand for blood transfusions. However, limitations like high costs and technological requirements restrict
Metal-mediated cross-linking in the generation of a marine-mussel adhesive.
Mary J Sever et al.
Angewandte Chemie (International ed. in English), 43(4), 448-450 (2004-01-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service