Skip to Content
Merck
All Photos(4)

Key Documents

C0755

Sigma-Aldrich

Conalbumin from chicken egg white

Substantially iron-free

Synonym(s):

Ovotransferrin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

chicken egg white

Assay

≥98% (agarose gel electrophoresis)

form

powder

technique(s)

protein quantification: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

chicken ... TF(396241)

Looking for similar products? Visit Product Comparison Guide

General description

Conalbumin is an iron-binding protein. It is mainly present in the egg white of hen. It is also termed as ovotransferrin. It is a member of the transferrin iron-binding glycoproteins family. Ovotransferrin is also known an an acute?phase protein (APP)in some organisms. It is the second main protein found in egg white.

Application

Conalbumin from chicken egg white has been used:
  • as ovotransferrin standard in 96?well microplate assay to investigate on ovotransferrin-like activity in the plasma of wild birds
  • to examine its effects on osteoclastogenesis
  • to study its partitioning in aqueous two-phase systems

Biochem/physiol Actions

Conalbumin/ovotransferrin plays a key role in avian natural immunity. It has the ability to prevent bacterial growth and possesses immunomodulatory functions. Ovotransferrin also exhibits antimicrobial and antioxidant activities.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siderophores and transferrins
Dertz EA and Raymond KN
ChemInform, 35(49), no-no (2004)
Miriam S Arrell et al.
Electrophoresis, 37(22), 2913-2921 (2016-10-21)
3-(2-furoyl)quinoline-2-carboxaldehyde (FQ) is a sensitive fluorogenic dye, used for derivatization of proteins for SDS-CGE with LIF detection (SDS-CGE-LIF) at silver staining sensitivity (ng/mL). FQ labels proteins at primary amines, found at lysines and N-termini, which vary in number and accessibility
Xiaowei Wu et al.
Journal of translational medicine, 19(1), 353-353 (2021-08-19)
Accumulating evidence highlights the critical roles of fibroblast growth factors (FGFs) in regulating the progression of multiple human cancers, including non-small cell lung cancer (NSCLC). In this study, we investigated the role of FGF11 in the progression of NSCLC. Previously
Innate immune responses
Avian Immunology, 121-147 (2014)
Long Huang et al.
Bioengineered, 13(1), 12-26 (2021-12-31)
Long non-coding RNAs (lncRNAs) are related to the initiation and progression of tumor and regulate various cellular processes including growth, invasion, migration, and apoptosis. Understanding the roles and mechanisms of lncRNAs in regulating cancer progression is crucial for formulating novel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service