Skip to Content
Merck
All Photos(2)

Documents

S7951

Sigma-Aldrich

Ovalbumin (257-264) chicken

≥97% (HPLC), lyophilized powder, antigenic peptide 

Synonym(s):

L-seryl-L-isoleucyl-L-isoleucyl-L-asparaginyl-L-phenylalanyl-L-a-glutamyl-L-lysyl- L-Leucine trifluoroacetate, OVA (257 - 264), SIINFEKL trifluoroacetate salt, Ser-Ile-Ile-Asn-Phe-Glu-Lys-Leu trifluoroaceate, pOV8

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H74N10O13 · xC2HF3O2
CAS Number:
Molecular Weight:
963.13 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Ovalbumin (257-264) chicken, ≥97% (HPLC)

Quality Level

Assay

≥97% (HPLC)

form

lyophilized powder

color

white to off-white

solubility

H2O: >1 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC[C@H](C)[C@H](NC(=O)[C@@H](N)CO)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(O)=O

InChI

1S/C45H74N10O13.C2HF3O2/c1-7-25(5)36(55-44(66)37(26(6)8-2)54-38(60)28(47)23-56)43(65)52-32(22-34(48)57)42(64)51-31(21-27-14-10-9-11-15-27)41(63)50-30(17-18-35(58)59)40(62)49-29(16-12-13-19-46)39(61)53-33(45(67)68)20-24(3)4;3-2(4,5)1(6)7/h9-11,14-15,24-26,28-33,36-37,56H,7-8,12-13,16-23,46-47H2,1-6H3,(H2,48,57)(H,49,62)(H,50,63)(H,51,64)(H,52,65)(H,53,61)(H,54,60)(H,55,66)(H,58,59)(H,67,68);(H,6,7)/t25-,26-,28-,29-,30-,31-,32-,33-,36-,37-;/m0./s1

InChI key

BRUCWCULSHVYBQ-VPZPQSBKSA-N

Amino Acid Sequence

Ser-Ile-Ile-Asn-Phe-Glu-Lys-Leu

Application

Ovalbumin (257-264) chicken has been used to trigger T cell activation in immunology studies.
Ovalbumin (257-264) chicken has been used:
  • to sensitize mice to generate an asthmatic model
  • in the stimulation of splenocytes
  • for treating dendritic cells to be used as a positive control in cross-presentation studies

Biochem/physiol Actions

Ovalbumin (257-264) chicken is an antigenic peptide and triggers immune response. Administration of the ovalbumin peptide in pregnant mice helps in surpassing ear development anomalies.
SIINFEKL is a class I (Kb)-restricted peptide epitope of ovalbumin (OVA); the ovalbumin fragment is presented by the class I MHC molecule, H-2Kb.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Primary in vivo responses to ovalbumin. Probing the predictive value of the Kb binding motif.
Lipford GB, et al.
Journal of Immunology, 150(4), 1212-1222 (1993)
Dead Tumor Cells Expressing Infectious Salmon Anemia Virus Fusogenic Protein Favor Antigen Cross-Priming In Vitro
Morales J, et al.
Frontiers in immunology, 8, 1170-1170 (2017)
Murine In vitro Memory T Cell Differentiation
Kim MV et al.
Bio-protocol, 4, 1-4 (2014)
Soluble peptide treatment reverses CD8 T-cell-induced disease in a mouse model of spontaneous tissue-selective autoimmunity
Paek SY, et al.
The Journal of Investigative Dermatology, 132(3), 677-686 (2012)
P-F Yu et al.
European review for medical and pharmacological sciences, 23(4), 1688-1697 (2019-03-07)
We aimed to investigate whether PM2.5 has the potential to exacerbate neutrophil airway inflammation and to analyze the underlying mechanisms. The high-volume air sampler (Laoying 2033B, Qingdao, China) was used to collect PM2.5 from January 01, 2016 to December 21

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service