Skip to Content
Merck
All Photos(4)

Documents

C2432

Sigma-Aldrich

Chloroform

contains 100-200 ppm amylenes as stabilizer, ≥99.5%

Synonym(s):

Methylidyne trichloride, Trichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38
Beilstein:
1731042
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.04

vapor density

4.1 (vs air)

Quality Level

vapor pressure

160 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

contains

100-200 ppm amylenes as stabilizer

technique(s)

RNA extraction: suitable

refractive index

n20/D 1.445 (lit.)

bp

60.5-61.5 °C (lit.)

mp

−63 °C (lit.)

density

1.492 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)Cl

InChI

1S/CHCl3/c2-1(3)4/h1H

InChI key

HEDRZPFGACZZDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroform is a colorless, volatile chlorinated organic solvent whose vapors have a narcotic effect. The product contains amylene as a stabilizer, since chloroform is susceptible to degradation with time. Amylene acts as a scavenger of free radicals formed during chloroform decomposition.

Application

Chloroform has been used for the isolation of RNA from mammalian tissue.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immune activation and target organ damage are consequences of hydrodynamic treatment but not delivery of naked siRNAs in mice.
Racz Z et al.
Nucleic Acid Therapeutics, 21, 215-215 (2011)
GC-MS-Based Analysis of Chloroform Extracted Suberin-Associated Root Waxes from Arabidopsis and Other Plant Species.
GC-MS-Based Analysis of Chloroform Extracted Suberin-Associated Root Waxes from Arabidopsis and Other Plant Species.
Kosma DK, et al.
Bio-protocol, 5(24) (2015)
Atrial fibrosis in a chronic murine model of obstructive sleep apnea: mechanisms and prevention by mesenchymal stem cells.
Ramos P et al.
Respiratory Research, 15, 54-54 (2014)
RNA Isolation from Synechocystis.
Mironov KS and. Los DA.
Bio-protocol, 5(6) (2015)
Min Wang et al.
Journal of colloid and interface science, 445, 84-92 (2015-01-18)
Phospholipids fulfill an important role in joint lubrication. They, together with hyaluronan and glycoproteins, are the biolubricants that sustain low friction between cartilage surfaces bathed in synovial fluid. In this work we have investigated how the friction force and load

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service