Skip to Content
Merck
All Photos(1)

Documents

B9935

Sigma-Aldrich

BMS-345541

≥98% (HPLC), powder

Synonym(s):

N-(1,8-Dimethylimidazo[1,2-a]quinoxalin-4-yl)-1,2-ethanediamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17N5 · HCl
CAS Number:
Molecular Weight:
291.78
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

solubility

DMSO: 18 mg/mL
H2O: insoluble

originator

Bristol-Myers Squibb

storage temp.

2-8°C

SMILES string

Cl[H].Cc1ccc2nc(NCCN)c3ncc(C)n3c2c1

InChI

1S/C14H17N5.ClH/c1-9-3-4-11-12(7-9)19-10(2)8-17-14(19)13(18-11)16-6-5-15;/h3-4,7-8H,5-6,15H2,1-2H3,(H,16,18);1H

InChI key

MIDKPVLYXNLFGZ-UHFFFAOYSA-N

Application

BMS-345541 has been used:
  • as an IκB kinase (IKK) complex inhibitor to study its effects on the interferon-γ (IFN-γ) production by natural killer (NK) cells
  • as an IKK inhibitor to study its effects on the expression of eotaxin and monocyte chemoattractant protein-1 (MCP-1) mRNA in gingival fibroblasts
  • as nuclear factor κB (NF-κB) pathway inhibitor to study its effects on tumor necrosis factor α (TNFα) production in human oral squamous carcinoma cells

Biochem/physiol Actions

BMS-345541 possesses anti-inflammatory properties. It also blocks the nuclear factor κB (NF-κB)-dependent transcription of pro-inflammatory cytokines. BMS-345541 possesses anti-inflammatory activity and plays a role in arresting bone erosion in certain animal models.
BMS-345541 is an IKB kinase (IKK) allosteric site inhibitor.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James R Burke et al.
The Journal of biological chemistry, 278(3), 1450-1456 (2002-10-31)
The signal-inducible phosphorylation of serines 32 and 36 of I kappa B alpha is critical in regulating the subsequent ubiquitination and proteolysis of I kappa B alpha, which then releases NF-kappa B to promote gene transcription. The multisubunit I kappa
Neety Sahu et al.
JCI insight, 7(20) (2022-10-05)
No disease-modifying drug exists for osteoarthritis (OA). Despite success in animal models, candidate drugs continue to fail in clinical trials owing to the unmapped interpatient heterogeneity and disease complexity. We used a single-cell platform based on cytometry by time-of-flight (cyTOF)
Christina Ward et al.
Tissue barriers, 3(1-2), e982424-e982424 (2015-04-04)
NF-κB (p50/p65) is the best characterized transcription factor known to regulate cell responses to inflammation. However, NF-κB is also constitutively expressed. We used inhibitors of the classical NF-κB signaling pathway to determine whether this transcription factor has a role in
Qinghai You et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 69(1), 75-85 (2019-11-07)
Acute respiratory distress syndrome (ARDS) is a life-threatening medical condition. It is characterized by serious lung inflammation or injury. Characterizing novel miRNAs implicated in ARDS pathogenesis may provide new therapeutic strategy for managing ARDS. We employed LPS-induced lung injury model
Feng He et al.
The Journal of experimental medicine, 218(3) (2020-12-15)
White adipose tissues (WAT) play crucial roles in maintaining whole-body energy homeostasis, and their dysfunction can contribute to hepatic insulin resistance and type 2 diabetes mellitus (T2DM). However, the mechanisms underlying these alterations remain unknown. By analyzing the transcriptome landscape

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service