Skip to Content
Merck
All Photos(2)

Documents

S8307

Sigma-Aldrich

SB 203580

≥98% (HPLC), solid, p38 MAPK inhibitor

Synonym(s):

4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16FN3OS
CAS Number:
Molecular Weight:
377.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

SB 203580, solid, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

−20°C

SMILES string

CS(=O)c1ccc(cc1)-c2nc(-c3ccc(F)cc3)c([nH]2)-c4ccncc4

InChI

1S/C21H16FN3OS/c1-27(26)18-8-4-16(5-9-18)21-24-19(14-2-6-17(22)7-3-14)20(25-21)15-10-12-23-13-11-15/h2-13H,1H3,(H,24,25)

InChI key

CDMGBJANTYXAIV-UHFFFAOYSA-N

Application

MDCK cells were treated with SB 203580 to study the role of MAPK signaling in inducing hypertonicity of cell membrane and potassium depletion.16 It was used to inhibit MAPK signaling in mouse cortical neurons17 and human hepatocellular carcinoma cell lines.18

Biochem/physiol Actions

SB 203580 is a pyridinyl imidazole that suppresses the activation of MAPKAP kinase-2 and inhibits the phosphorylation of heat shock protein (HSP) 27 in response to IL-1, cellular stresses and bacterial endotoxin in vivo. It does not inhibit JNK or p42 MAP kinase and therefore, is useful for studying the physiological roles and targets of p38 MAPK and MAPKAP kinase-2. It has been shown to induce the activation of the serine/threonine kinase Raf-1 and has been reported to inhibit cytokine production.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harini Lakshminarasimhan et al.
Scientific reports, 7(1), 4533-4533 (2017-07-05)
Doxorubicin (DOX), a common chemotherapeutic agent, impairs synaptic plasticity. DOX also causes a persistent increase in basal neuronal excitability, which occludes serotonin-induced enhanced excitability. Therefore, we sought to characterize and reverse DOX-induced physiological changes and modulation of molecules implicated in
S Hashimoto et al.
Biochemical and biophysical research communications, 258(2), 443-447 (1999-05-18)
We examined the regulatory role of a reduction/oxidation (redox) control protein, thioredoxin (TRX), in tumor necrosis factor-alpha (TNF-alpha)-induced p38 MAP kinase activation and p38 MAP kinase-mediated cytokine expression utilizing TRX-transfected murine L929 cells (TRX14). The results showed that TNF-alpha-induced p38
Jianxin Huo et al.
Cells, 9(3) (2020-03-04)
: Apoptosis signal-regulating kinase 1 (ASK1) is a mitogen-activated protein kinase kinase kinase (MAPKKK) that activates downstream JNK and p38 mitogen-activated protein kinase (MAPK) to relay death signals into cells in response to various environmental stress. However, whether ASK1 plays
Ying Yang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 42(5), 2130-2143 (2017-08-16)
C1q and tumour necrosis factor-related protein 1 (CTRP1) possesses anti-atherogenic and anti-inflammatory effects. This study investigated whether the CTRP1 levels in the plasma and epicardial adipose tissue (EAT) were associated with congestive heart failure (CHF) and to disclose possible molecular
Camille Robinet et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 30(2), 286-298 (2009-10-02)
MCT2 is the predominant neuronal monocarboxylate transporter allowing lactate use as an alternative energy substrate. It is suggested that MCT2 is upregulated to meet enhanced energy demands after modifications in synaptic transmission. Brain-derived neurotrophic factor (BDNF), a promoter of synaptic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service