Skip to Content
Merck
All Photos(4)

Key Documents

B4679

Sigma-Aldrich

BIS-TRIS propane

BioPerformance Certified, suitable for cell culture, ≥99.0%

Synonym(s):

1,3-Bis[tris(hydroxymethyl)methylamino]propane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[CH2NHC(CH2OH)3]2
CAS Number:
Molecular Weight:
282.33
Beilstein:
1786109
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

BioPerformance Certified

Quality Level

Assay

≥99.0%

form

crystals

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin and bioburden, tested

useful pH range

6.3-9.5

pKa (25 °C)

(1) 6.8, (2) 9.0

mp

164-165 °C (lit.)

application(s)

diagnostic assay manufacturing

foreign activity

DNAse (exonuclease), NICKase (endonuclease), RNAse, and protease, none detected

SMILES string

OCC(CO)(CO)NCCCNC(CO)(CO)CO

InChI

1S/C11H26N2O6/c14-4-10(5-15,6-16)12-2-1-3-13-11(7-17,8-18)9-19/h12-19H,1-9H2

InChI key

HHKZCCWKTZRCCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Neutralized chimeric DNA probe for the improvement of GC-rich RNA detection specificity on the nanowire field-effect transistor.: BIS-TRIS propane is used in the development of nanowire field-effect transistors for detecting GC-rich RNA sequences. The study demonstrates the buffer′s role in enhancing the specificity and sensitivity of these biosensors (Chou et al., 2019).

  • Elimination of ghost peaks by optimization of anion exchange chromatography method for determination of gamma-carboxyglutamic acid (Gla)-domainless impurity in recombinant activated clotting factor VII drug products.: BIS-TRIS propane is employed in optimizing chromatography methods to eliminate ghost peaks during the analysis of pharmaceutical impurities. The buffer′s effectiveness ensures accurate and reliable detection of impurities in drug formulations (Tahmasbi et al., 2019).

Other Notes

Easily compare specifications for BIS-TRIS propane products with the BIS-TRIS propane specification table.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph S Brunzelle et al.
Archives of biochemistry and biophysics, 474(1), 157-166 (2008-04-01)
The three-dimensional structure of the catalytically efficient beta-xylosidase from Selenomonas ruminantium in complex with competitive inhibitor 1,3-bis[tris(hydroxymethyl)methylamino]propane (BTP) was determined by using X-ray crystallography (1.3A resolution). Most H bonds between inhibitor and protein occur within subsite -1, including one between
Sami J O Varjo et al.
Journal of chromatography. A, 1081(1), 92-98 (2005-07-15)
Greater stability of liposome coatings and improved resolution of model steroids in capillary electrochromatography (CEC) were sought by adding small diamines (ethylenediamine, diaminopropane, bis-tris-propane, or N-(2-hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid, HEPES)) to the liposome solution before coating of fused silica capillaries. The phospholipid
S J Smith et al.
The Journal of biological chemistry, 259(15), 9365-9368 (1984-08-10)
The kinetics of binding and hydrolysis of ATP by bovine cardiac myosin subfragment 1 has been reinvestigated. More than 90% of the total fluorescence amplitude associated with ATP hydrolysis occurs with an apparent second-order rate constant of 8.1 X 10(5)
M H Hopkins et al.
Journal of enzyme inhibition, 6(3), 195-199 (1992-01-01)
It is hypothesized that buffers capable of forming a Schiff base with the PLP of gamma-aminobutyric acid aminotransferase (GABA-AT) may lead to denaturation and inactivation of the enzyme. On the basis of this hypothesis three new methods for the selective
E Csuhai et al.
Biochemistry, 34(38), 12411-12419 (1995-09-26)
A secreted dibasic cleaving peptidase capable of converting dynorphins into Leu-enkephalin-Arg6 was purified from the medium of EL-4 mouse thymoma cells. The enzyme is a novel metalloendopeptidase with a neutral pH optimum (6.9) and a molecular weight of approximately 130

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service