Skip to Content
Merck
All Photos(1)

Key Documents

B1686

Sigma-Aldrich

BIO

≥98% (HPLC), powder, GSK-3α/β inhibitor

Synonym(s):

(2’Z,3’E)-6-Bromoindirubin-3′-oxime, 6-BIO, 6BIO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10BrN3O2
CAS Number:
Molecular Weight:
356.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

BIO, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

dark red

solubility

DMSO: >5 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O\N=C1\C(Nc2ccccc12)=C3\C(=O)Nc4cc(Br)ccc34

InChI

1S/C16H10BrN3O2/c17-8-5-6-9-12(7-8)19-16(21)13(9)15-14(20-22)10-3-1-2-4-11(10)18-15/h1-7,18,22H,(H,19,21)/b15-13-,20-14+

InChI key

DDLZLOKCJHBUHD-WAVHTBQISA-N

Application

6-bromoindirubin-3′-oxime (BIO) has been used:
  • in MTT 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl tetrazolium bromide) cell proliferation assay
  • as a medium supplement in embryonic stem cells (ESCs)
  • for the inhibition of glycogen synthase kinase 3 β (GSK-3β) in human dermal papilla cells (hDPCs)

Biochem/physiol Actions

6-bromoindirubin-3′-oxime (BIO) is a potent, reversible and ATP-competitive GSK-3α/β inhibitor and the first pharmacological agent shown to maintain self-renewal in human and mouse embryonic stem cells. Human embryonic stem cells (hESCs) are maintained in the undifferentiated state through treatment with a GSK-3 inhibitor, BIO, under a feeder-free condition.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GSK-3 page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

air sensitive

Legal Information

Product is sold for research and laboratory purposes in vitro and may not be used for any in vivo or therapeutic uses in humans, veterinary applications, or to make chemical derivatives.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew R Patterson et al.
Nature communications, 9(1), 430-430 (2018-02-01)
GTPase of immunity-associated protein 5 (Gimap5) is linked with lymphocyte survival, autoimmunity, and colitis, but its mechanisms of action are unclear. Here, we show that Gimap5 is essential for the inactivation of glycogen synthase kinase-3β (GSK3β) following T cell activation.
Human placental extract exerts hair growth-promoting effects through the GSK-3beta signaling pathway in human dermal papilla cells
Kwon TR, et al.
International Journal of Molecular Medicine, 36(4), 1088-1096 (2015)
Tatsuki Ogasawara et al.
Scientific reports, 7(1), 8630-8630 (2017-08-19)
Pluripotent stem cell-derived cardiomyocytes show great promise in regenerating the heart after myocardial infarction; however, several uncertainties exist that must be addressed before clinical trials. One practical issue is graft survival following transplantation. Although a pro-survival cocktail with Matrigel has
Xiang Qiu et al.
World journal of gastroenterology, 21(26), 8061-8072 (2015-07-18)
To investigate the therapeutic effects of lutein against non-alcoholic fatty liver disease (NAFLD) and the related underlying mechanism. After 9 d of acclimation to a constant temperature-controlled room (20 °C-22 °C) under 12 h light/dark cycles, male Sprague-Darley rats were randomly divided
Noëmie Jean LeBlanc et al.
Molecular neurobiology, 56(9), 6521-6538 (2019-03-11)
Stroke induces blood-brain barrier (BBB) breakdown, which promotes complications like oedema and hemorrhagic transformation. Administration of recombinant tissue plasminogen activator (rtPA) within a therapeutic time window of 4.5 h after stroke onset constitutes the only existing treatment. Beyond this time window

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service