Skip to Content
Merck
All Photos(1)

Key Documents

368050

Sigma-Aldrich

Granzyme B Inhibitor I

≥95% (HPLC), solid, Granzyme B inhibitor, Calbiochem®

Synonym(s):

Granzyme B Inhibitor I, Z-AAD-CMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24ClN3O7
Molecular Weight:
441.86
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Granzyme B Inhibitor I, The Granzyme B Inhibitor I controls the biological activity of Granzyme B. This small molecule/inhibitor is primarily used for Cancer applications.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A weak inhibitor of the human and murine granzyme B. Also inhibits the apoptosis-related DNA fragmentation in lymphocytes by fragmentin 2, a rat lymphocyte granule protease homologous to granzyme B (ID50 = 300 nM).
A weak inhibitor of the human and murine serine protease granzyme B. Also inhibits the apoptosis related DNA fragmentation in lymphocytes by fragmentin 2, a rat lymphocyte granule protease homologous to granzyme B (ID50 = 300 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Granzyme B
Product does not compete with ATP.
Reversible: no
Target IC50: 300 nM against the apoptosis-related DNA fragmentation in lymphocytes by fragmentin 2

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Ala-Ala-Asp-CH₂Cl

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gong, B., et al. 1999. Cell Growth Different. 10, 491.
Shi, L., et al. 1992. J. Exp. Med. 176, 1521.
Odake, S., et al. 1991. Biochemistry30, 2217.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ganta Vijay Chaitanya et al.
Brain pathology (Zurich, Switzerland), 21(1), 16-30 (2010-09-10)
Infiltration of leukocytes into post-ischemic cerebrum is a well-described phenomenon in stroke injury. Because CD-8(+) T-lymphocytes secrete cytotoxic proteases, including granzyme-b (Gra-b) that exacerbates post-ischemic brain damage, we investigated roles of Gra-b in human stroke. To study the role of
Ning-Ning Ji et al.
Frontiers in neurology, 10, 1306-1306 (2020-01-11)
Hippocampal neuronal apoptosis is a devastating consequence of cardiac arrest (CA) and subsequent cardiopulmonary resuscitation (CPR). In this study, we assessed the contribution of cytotoxic T lymphocyte (CTL)-derived toxic mediator granzyme B (Gra-b) to the hippocampal neuronal apoptosis following CA/CPR
Yan Su et al.
Cell discovery, 8(1), 35-35 (2022-04-20)
Penetration of immune cells into tumor cells was believed to be immune-suppressive via cell-in-cell (CIC) mediated death of the internalized immune cells. We unexpectedly found that CIC formation largely led to the death of the host tumor cells, but not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service