Skip to Content
Merck
All Photos(1)

Key Documents

A9887

Sigma-Aldrich

Aflatoxin B2

≥98% (HPLC), powder, mycotoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O6
CAS Number:
Molecular Weight:
314.29
Beilstein:
1355115
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Aflatoxin B2,

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

COc1cc2O[C@H]3OCC[C@H]3c2c4OC(=O)C5=C(CCC5=O)c14

InChI

1S/C17H14O6/c1-20-10-6-11-14(8-4-5-21-17(8)22-11)15-13(10)7-2-3-9(18)12(7)16(19)23-15/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1

InChI key

WWSYXEZEXMQWHT-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Aflatoxin B2/AFB2 is a toxic mycotoxin.

Application

Aflatoxin B2 has been used as a standard in a study to find out aflatoxins in various foods, spices and feedstuffs by high performance liquid chromatography (HPLC). It has also been used as a standard to determine aflatoxins in honey by HPLC.

Biochem/physiol Actions

Aflatoxin B2 is a hepatocarcinogen. Food contaminant produced by Aspergillus flavus, a common soil fungus. Aflatoxin B2/AFB2 may have dangerous hepatotoxic, teratogenic and carcinogenic actions on various forms of livestock.

Warning

Possibly carcinogenic.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Carc. 1B - Muta. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A STUDY ON AFLATOXINS IN VARIOUS FOODS, SPICES AND FEEDSTUFFS BY HIGH PERFORMANCE LIQUID CHROMATOGRAPHY
OZ, T. Y. V.
The ISME Journal, 31, 11-16 (2016)
Induction of mitochondria-mediated apoptosis and PI3K/Akt/mTOR-mediated autophagy by aflatoxin B2 in hepatocytes of broilers
Chen B, et al.
Oncotarget, 7(51), 84989-84989 (2016)
Bacillaceae spores, fungi and aflatoxins determination in honey Esporos de Bacillaceae, fungos e aflatoxinas em mel
Martins H M, et al.
RN (2003)
Fernanda Cosme et al.
Foods (Basel, Switzerland), 9(12) (2020-12-06)
Aflatoxins B1 and B2 are two highly toxic mycotoxins that have been sometimes found in wines. Currently, no technological solution is available to reduce or eliminate aflatoxins from wines when they are present. Therefore, this work aims to study the
Sharaf S Omar et al.
Foods (Basel, Switzerland), 9(5) (2020-05-24)
In Jordan as in other worldwide countries, mycotoxins are considered a serious national problem in food supplies. As a result, almost all nations are setting and adopting different regulations targeting the control of mycotoxins levels in the domestic food supply

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service