Skip to Content
Merck
All Photos(1)

Key Documents

34029

Supelco

Aflatoxin B1 solution

2 μg/mL in acetonitrile, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12O6
CAS Number:
Molecular Weight:
312.27
Beilstein:
1269174
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

2 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

[H][C@]12OC=C[C@@]1([H])c3c(O2)cc(OC)c4C5=C(C(=O)CC5)C(=O)Oc34

InChI

1S/C17H12O6/c1-20-10-6-11-14(8-4-5-21-17(8)22-11)15-13(10)7-2-3-9(18)12(7)16(19)23-15/h4-6,8,17H,2-3H2,1H3/t8-,17+/m0/s1

InChI key

OQIQSTLJSLGHID-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aflatoxin B1 is a mycotoxin found to be hepatocarcinogenic in nature. It has a strong affinity for the deoxyribonucleic acid (DNA) and binds to both native and denatured deoxyribonucleic acid (DNA) as indicated by spectroscopy and equilibrium dialysis results. This fungal metabolic product is commonly found in food and feed, contaminated by Aspergillus species.

The product is supplied in CERTAN® vial.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Aflatoxin B1 solution may be used as an analytical reference standard for the determination of the analyte in meat, meat products, eggs and milk using high-performance liquid chromatography (HPLC) with fluorescence and ultraviolet detection.

Legal Information

CERTAN is a registered trademark of LGC Group

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susceptibility to hepatocellular carcinoma is associated with genetic variation in the enzymatic detoxification of aflatoxin B1.
McGlynn AK, et al.
Proceedings of the National Academy of Sciences of the USA, 92(6), 2384-2387 (1995)
Aflatoxin B1 induces the transversion of G-> T in codon 249 of the p53 tumor suppressor gene in human hepatocytes.
Aguilar F, et al.
Proceedings of the National Academy of Sciences of the USA, 90(18), 8586-8590 (1993)
Anna Z Wec et al.
Cell host & microbe, 25(1), 39-48 (2019-01-11)
Passive administration of monoclonal antibodies (mAbs) is a promising therapeutic approach for Ebola virus disease (EVD). However, all mAbs and mAb cocktails that have entered clinical development are specific for a single member of the Ebolavirus genus, Ebola virus (EBOV)
Determination of aflatoxins in eggs, milk, meat and meat products using HPLC fluorescent and UV detectors.
Herzallah MS.
Food Chemistry, 114(3), 1141-1146 (2009)
M B Sporn et al.
Science (New York, N.Y.), 151(3717), 1539-1541 (1966-03-25)
Aflatoxin B(1) binds to both native and denatured DNA, as shown by spectroscopy and equilibrium dialysis. It also strongly inhibits incorporation of cytidine into rat liver nuclear RNA and lowers the RNA content of the nucleus. The extremle toxicity and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service