Skip to Content
Merck
All Photos(3)

Key Documents

A6636

Sigma-Aldrich

Aflatoxin B₁

greener alternative

from Aspergillus flavus, ≥98.0% (HPLC), powder, CYP1A2, CYP2A6, CYP2D6, and CYP3A family substrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12O6
CAS Number:
Molecular Weight:
312.27
Beilstein:
1269174
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus

biological source

Aspergillus flavus

Quality Level

Assay

≥98.0% (HPLC)

form

powder

greener alternative product score

old score: 51
new score: 42
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

SMILES string

COc1cc2OC3OC=CC3c2c4OC(=O)C5=C(CCC5=O)c14

InChI

1S/C17H12O6/c1-20-10-6-11-14(8-4-5-21-17(8)22-11)15-13(10)7-2-3-9(18)12(7)16(19)23-15/h4-6,8,17H,2-3H2,1H3/t8-,17+/m0/s1

InChI key

OQIQSTLJSLGHID-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Designing Safer Chemicals” and “Design for Energy Efficiency”. Click here to view its DOZN scorecard.

Application

Aflatoxin B1 is a carcinogenic compound that induces transversion of G to T at codon 249 of the p53 tumor suppressor gene. Aflatoxin B1 may be used as an internal standard when testing for aflatoxin contamination in food products.

Biochem/physiol Actions

Aflatoxin B1 is a carcinogenic compound produced by Aspergillus flavus, a common soil fungus, that induces transversion of G to T at codon 249 of the p53 tumor suppressor gene. Aflatoxin B1 is a food contaminant and a hepatocarcinogen. Aflatoxin is biotransformed to genotoxic intermediates by P450 Phase I enzymes, mainly CYP3A4 via aflatoxin B1 3-hydroxylation. Detoxification depends on Phase II enzymes, such as Glutathione S-Transferase and AFB(1)-aldehyde reductase (AFAR). Aflatoxin B1 is a CYP1A2, CYP2A6, CYP2D6, and CYP3A family substrate.

Warning

Possibly carcinogenic.

standard

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Carc. 1B - Muta. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hajar Zolfaghari et al.
Advanced pharmaceutical bulletin, 10(3), 482-487 (2020-07-16)
Purpose: The present study was conducted to assess the ability of probiotic bacteria and yeasts strains to reduce aflatoxin B1 (AFB1) in gastrointestinal simulated conditions. Aflatoxins are potent carcinogenic and immunosuppressive agents. Acute exposure to a high level of aflatoxins
Rahul R Bhowate et al.
Journal of oral and maxillofacial pathology : JOMFP, 25(1), 110-117 (2021-08-06)
Areca nut is deeply rooted sociocultural habit in India. Areca nut reported to be infested by fungi during the field and storage conditions. Areca nut alkaloids, nitrosamines, tobacco and aflatoxin are cytotoxic, immunotoxic to red blood cell and epithelial cell.
Andre Barany et al.
Animals : an open access journal from MDPI, 11(3) (2021-04-04)
Several studies in fish have shown that aflatoxin B1 (AFB1) causes a disparity of species-dependent physiological disorders without compromising survival. We studied the effect of dietary administration of AFB1 (2 mg AFB1 kg-1 diet) in gilthead seabream (Sparus aurata) juveniles
Ilenia Siciliano et al.
Toxins, 7(3), 743-754 (2015-03-10)
Fungi belonging to the genus Alternaria are common pathogens of fruit and vegetables with some species able to produce secondary metabolites dangerous to human health. Twenty-eight Alternaria isolates from rocket and cabbage were investigated for their mycotoxin production. Five different
Agnieszka Chlebicz et al.
Probiotics and antimicrobial proteins, 12(1), 289-301 (2019-02-06)
The aim of the following research was to determine the detoxification properties of probiotic Lactobacillus sp. bacteria (12 strains) and S. cerevisiae yeast (6 strains) towards mycotoxins, such as aflatoxin B1, deoxynivalenol, fumonisins, T-2 toxin and zearalenone, which pose as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service