Skip to Content
Merck
All Photos(1)

Key Documents

D0156

Sigma-Aldrich

Deoxynivalenol

Synonym(s):

3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, Vomitoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O6
CAS Number:
Molecular Weight:
296.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

[H][C@]12O[C@]3([H])[C@H](O)C[C@@](C)([C@]34CO4)[C@@]1(CO)[C@H](O)C(=O)C(C)=C2

InChI

1S/C15H20O6/c1-7-3-9-14(5-16,11(19)10(7)18)13(2)4-8(17)12(21-9)15(13)6-20-15/h3,8-9,11-12,16-17,19H,4-6H2,1-2H3/t8-,9-,11-,12-,13-,14-,15+/m1/s1

InChI key

LINOMUASTDIRTM-QGRHZQQGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Deoxynivalenol (DON) is a natural mycotoxin found in Fusarium graminearum and Fusarium culmorum. It belongs to the B-trichothecenes family of mycotoxins.

Application

Deoxynivalenol has been used:

  • as a mycotoxin to induce cytotoxicity in porcine jejunal epithelial cells and study the protective effects of Saccharomyces cerevisiae on the cell viability of host cells.
  • to induce toxicity in nude mice to study the effect of probiotics (Lactobacillus rhamnosus GG (LGG) or Lactobacillus acidophilus (LA)) supplementation on host protection
  • as a thyroid hormone receptor (TRβ) antagonist in cell-based luciferase reporter gene (CALUX) assay

Biochem/physiol Actions

Deoxynivalenol (DON) blocks the synthesis of DNA and RNA by inhibiting the process of translation.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxicology of deoxynivalenol (vomitoxin).
Rotter B A.
Int J Toxicol Pharmacol Res, 48(1), 1-34 (1996)
Yang Liu et al.
AMB Express, 9(1), 137-137 (2019-09-05)
Deoxynivalenol (DON) is one of the mycotoxins most frequently encountering in cereal-based foods throughout the world. Saccharomyces cerevisiae was used to alleviate porcine jejunal epithelia cell (IPEC-J2) injury induced by DON in this study. The results indicated that cell viability
Jun-Lin Song et al.
Cells, 10(8) (2021-08-28)
(1) Background: Deoxynivalenol (DON) and zearalenone (ZEA) are type B trichothecene mycotoxins that exert serious toxic effects on the reproduction of domestic animals. However, there is little information about the toxicity of mycotoxins on testis development in Equus asinus. This
Deoxynivalenol and its toxicity
Sobrova P, et al.
Interdisciplinary Sciences, Computational Life Sciences, 3(3), 94-99 (2010)
Determination of deoxynivalenol (DON) and its derivatives: Current status of analytical methods
Ran R, et al.
Food Control, 34(1), 138-148 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service