Skip to Content
Merck
All Photos(2)

Key Documents

471933

Sigma-Aldrich

4-Iodophenylboronic acid

≥95.0%

Synonym(s):

B-(4-iodophenyl)-boronic acid, p-Iodophenylboronic acid, p-iodo-benzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4B(OH)2
CAS Number:
Molecular Weight:
247.83
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0%

mp

326-330 °C (lit.)

functional group

iodo

SMILES string

OB(O)c1ccc(I)cc1

InChI

1S/C6H6BIO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4,9-10H

InChI key

PELJYVULHLKXFF-UHFFFAOYSA-N

Application

Reagent used for
  • Copper-mediated ligandless aerobic fluoroalkylation
  • Palladium-catalyzed aerobic oxidative cross-coupling reactions
  • Recyclable magnetic-nanoparticle-supported palladium catalyst for the Suzuki coupling reactions
  • Oxidative hydroxylation using a copper (Cu) catalyst
  • Ligand-free palladium-catalyzed Suzuki-Miyaura cross-coupling
  • Homocoupling using gold salts as a catalyst
  • Ruthenium (Ru)-catalyzed cross-coupling
  • CuI-catalyzed Suzuki coupling reactions
  • Palladium-catalyzed domino Heck-Mizoroki/Suzuki-Miyaura reactions
  • Manganese triacetate-mediated radical additions of arylboronic acids to alkenes

Reagent used in Preparation of
  • Pleuromutilin derivatives for ribosomal binding and antibacterial activity via "Click Chemistry"
  • Liquid crystalline polyacetylene derivatives

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CuI-catalyzed Suzuki coupling reaction of organoboronic acids with alkynyl bromides
S. Wang, et al.,
Tetrahedron, 67, 4800-4806 (2011)
B M Kinsey et al.
Nuclear medicine and biology, 20(1), 13-22 (1993-01-01)
Three iodinated phenylboronic acids have been synthesized: 4-iodophenylboronic acid (2a), 3-(4-iodobenzenesulfonamido)phenylboronic acid (5a) and 3-(5-dimethylamino-6-iodo-1-naphthalenesulfonamido)phenylboronic acid (6a). The corresponding no-carrier-added 125I derivatives 2b, 5b and 6b have been prepared in good yield by selective displacement of the tributylstannyl group. Compound
Kiyofumi Inamoto et al.
Chemical communications (Cambridge, England), 47(42), 11775-11777 (2011-10-01)
Copper-catalysed oxidative hydroxylation of arylboronic acids was accomplished in water containing an amphiphilic surfactant, providing facile access to phenol derivatives.
Disubstituted Liquid Crystalline Polyacetylene Derivatives That Exhibit Linearly Polarized Blue and Green Emissions
San Jose, B. A.; et al.
Macromolecules, 44, 6288-6302 (2011)
Alison M Berezuk et al.
Scientific reports, 8(1), 12933-12933 (2018-08-30)
In Escherichia coli, formation of new cells is mediated by the elongasome and divisome that govern cell elongation and septation, respectively. Proper transition between these events is essential to ensure viable progeny are produced; however, the components of each complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service