Skip to Content
Merck

Skip To

481908

FK866 hydrochloride hydrate

The Nicotinamide Phosphoribosyltransferase Inhibitor, FK866, also referenced under CAS 658084-64-1, controls the biological activity of Nicotinamide Phosphoribosyltransferase. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

Nicotinamide Phosphoribosyltransferase Inhibitor, FK866, (E)-N-(4-(1-Benzoylpiperidin-4-yl)butyl)-3-(pyridin-3-yl)acrylamide, NAMPT Inhibitor, APO866, NAMPTase Inhibitor I, PBEF Inhibitor I, Visfatin Inhibitor I, NAPRT Inhibitor, APO866, NAMPTase Inhibitor I, PBEF Inhibitor I, Visfatin Inhibitor I, NAPRT Inhibitor, (E)-N-(4-(1-Benzoylpiperidin-4-yl)butyl)-3-(pyridin-3-yl)acrylamide, NAMPT Inhibitor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H29N3O2
CAS Number:
Molecular Weight:
391.51
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Assay:
≥97% (HPLC)
Form:
liquid
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Nicotinamide Phosphoribosyltransferase Inhibitor, FK866, The Nicotinamide Phosphoribosyltransferase Inhibitor, FK866, also referenced under CAS 658084-64-1, controls the biological activity of Nicotinamide Phosphoribosyltransferase. This small molecule/inhibitor is primarily used for Neuroscience applications.

Quality Segment

assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N2(CCC(CC2)CCCCNC(=O)\C=C\c3cnccc3)C(=O)c1ccccc1

InChI

1S/C24H29N3O2/c28-23(12-11-21-8-6-15-25-19-21)26-16-5-4-7-20-13-17-27(18-14-20)24(29)22-9-2-1-3-10-22/h1-3,6,8-12,15,19-20H,4-5,7,13-14,16-18H2,(H,26,28)/b12-11+

InChI key

KPBNHDGDUADAGP-VAWYXSNFSA-N

General description

A cell-permeable pyridinylacrylamide compound that acts as a selective, allosteric NAPRT/NAMPT (nicotinamide phosphoribosyltransferase) inhibitor (Ki = 0.4 nM for the enzyme/substrate complex; Ki = 0.3 nM for the free enzyme), while exhibiting no effect toward NPRT (nicotinic acid phosphoribosyltransferase) activity (82% inhibition of NAPRT activity at 10 nM vs no inhibition of NPRT activity at 1 µM in K-562 extract). FK866 treatment is shown to induce cell death by depleting NAD+ (Cat. Nos. 481911 & 481915 ) in HepG2 and NIH-3T3 cultures (by >95% after 24 h treatment of 10 nM FK866), likewise the addition of exogenous NAD+ is demonstrated to rescue NIH-3T3 and SH-SY5Y from FK866-induced NAD+-depletion and cell death.

Packaging

Packaged under inert gas

Physical form

Supplied as a 50 mM (2 mg/102.17 µl) solution in DMSO.

Other Notes

Formentini, L., et al. 2009. Biochem. Pharmacol.77, 1612.
Nakahata, Y., et al. 2009. Science324, 654.
Ramsey. K.M., et al. 2009. Science324, 651.
Billington, R.A., et al. 2008. J. Biol. Chem.283, 6367.
Hasmann, M. and Schemainda, I., 2003. Cancer Res.63, 7436.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
480073406170171261
assay

≥97% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

form

liquid

form

solid

form

solid

form

liquid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, desiccated (hygroscopic), protect from light


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters