Skip to Content
Merck
All Photos(2)

Key Documents

A7902

Sigma-Aldrich

2-Amino-2-norbornanecarboxylic acid

suitable for ligand binding assays

Synonym(s):

BCH, 2-Aminobicyclo[2.2.1]heptane-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13NO2
CAS Number:
Molecular Weight:
155.19
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

product name

2-Amino-2-norbornanecarboxylic acid, amino acid transport inhibitor

Quality Level

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

mp

>300 °C (lit.)

SMILES string

NC1(C[C@@H]2CC[C@H]1C2)C(O)=O

InChI

1S/C8H13NO2/c9-8(7(10)11)4-5-1-2-6(8)3-5/h5-6H,1-4,9H2,(H,10,11)/t5-,6+,8?/m1/s1

InChI key

MPUVBVXDFRDIPT-RSHNMJPRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Amino-2-norbornanecarboxylic acid is a potential large amino acid transporters inhibitor. It may suppress tumor progression and apoptosis.

Application


  • Thyroid Hormone Transporters in a Human Placental Cell Model.: This study investigates the role of 2-Amino-2-norbornanecarboxylic acid in the transport of thyroid hormones in placental cells, offering insights that could improve understanding of fetal development and maternal health (Chen et al., 2022).

  • Metabolic adaptations to hypoxia in the neonatal mouse forebrain can occur independently of the transporters SLC7A5 and SLC3A2.: Research explores how 2-Amino-2-norbornanecarboxylic acid affects metabolic responses to hypoxia in neonatal brain development, providing a foundation for future studies on brain health and neurodevelopment (Fitzgerald et al., 2021).

  • Hemocompatible LAT1-inhibitor can induce apoptosis in cancer cells without affecting brain amino acid homeostasis.: This article assesses the potential of 2-Amino-2-norbornanecarboxylic acid as a selective inhibitor that could lead to new treatments for cancer while preserving critical brain functions (Markowicz-Piasecka et al., 2020).

  • Regulation of Melanogenesis by the Amino Acid Transporter SLC7A5.: Research demonstrates the utility of 2-Amino-2-norbornanecarboxylic acid in regulating skin pigmentation processes, which could have implications for disorders related to pigmentation (Gaudel et al., 2020).

  • Structure of the human LAT1-4F2hc heteromeric amino acid transporter complex.: This study utilizes 2-Amino-2-norbornanecarboxylic acid to elucidate the structure of a key amino acid transporter, which is crucial for understanding nutrient uptake and its implications in various diseases (Yan et al., 2019).


Biochem/physiol Actions

2-Amino-2-norbornanecarboxylic acid (BCH) is a Na(+)-independent system L specific substrate used with other system specific substrates such as the system A specific substrate α-(methylamino) isobutyric acid and system N specific substrate L-glutamic acid gamma-monohydroxamate to help identify, differentiate and characterize amino acid transport systems in cells.

Quality

Mixture of exo- and endo-isomers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Discovery of Potent Inhibitors for the Large Neutral Amino Acid Transporter 1 (LAT1) by Structure-Based Methods
Singh N, et al.
International Journal of Molecular Sciences, 20(1), 27-27 (2019)
Hiroyuki Okudaira et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 52(5), 822-829 (2011-05-04)
Trans-1-amino-3-(18)F-fluorocyclobutanecarboxylic acid (anti-(18)F-FACBC) is an amino acid PET tracer that has shown promise for visualizing prostate cancer. Therefore, we aimed to clarify the anti-(18)F-FACBC transport mechanism in prostate cancer cells. We also studied the fate of anti-(18)F-FACBC after it is
Lili Quan et al.
Journal of experimental & clinical cancer research : CR, 39(1), 266-266 (2020-12-02)
Tumor angiogenesis is regarded as a rational anti-cancer target. The efficacy and indications of anti-angiogenic therapies in clinical practice, however, are relatively limited. Therefore, there still exists a demand for revealing the distinct characteristics of tumor endothelium that is crucial
Kohichi Yamauchi et al.
Cancer letters, 276(1), 95-101 (2008-12-09)
LAT1, a subunit of heterodimeric system L transporter responsible for transporting neutral amino acids into cells, has been investigated in several cancers because of its onco-fetal nature. Based on the studies of its functional inhibition, LAT1 has been proposed to
M M Sánchez del Pino et al.
The Journal of biological chemistry, 267(36), 25951-25957 (1992-12-25)
Endothelial cell membranes, the site of the blood-brain barrier, were obtained from the capillaries of cow brain. The luminal and abluminal membranes were separated by centrifugation on a discontinuous Ficoll gradient. Electron microscopy revealed that the membrane preparations consisted almost

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service