Skip to Content
Merck
All Photos(1)

Key Documents

M2383

Sigma-Aldrich

α-(Methylamino)isobutyric acid

≥97% (titration)

Synonym(s):

2,N-Dimethylalanine, 2-(Methylamino)-2-methylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHC(CH3)2COOH
CAS Number:
Molecular Weight:
117.15
Beilstein:
1746984
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥97% (titration)

form

powder

technique(s)

ligand binding assay: suitable

color

white

mp

>300 °C (lit.)

storage temp.

−20°C

SMILES string

CNC(C)(C)C(O)=O

InChI

1S/C5H11NO2/c1-5(2,6-3)4(7)8/h6H,1-3H3,(H,7,8)

InChI key

DLAMVQGYEVKIRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

α-(Methylamino)isobutyric acid (MeAIB) has been used:
  • as a system A transport system inhibitor to perform inhibition experiments
  • as a system A transport system inhibitor to study its effect on the generation of epileptiform field potentials (EFPs) in the acutely disinhibited cortical slices
  • as an N-acetyltransferase (SNAT) inhibitor in Hank′s balanced salt solution (HBSS) to study its influence on the uptake of 14C-cysteine/ radioactive-labeled cysteine

Biochem/physiol Actions

α-(Methylamino)isobutyric acid (MeAIB), a competitive inhibitor of the neutral amino acid transport A system, is an N-methylated substrate. It is capable of decreasing the amplitude of the miniature excitatory postsynaptic current (mEPSC) in hippocampus neurons developing on top of astrocytes. MeAIB can block the alanine-proline (AP) pathway.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Pajor et al.
The Journal of membrane biology, 107(3), 237-247 (1989-03-01)
Brush border membrane vesicles (BBMV) were prepared from the gills of the marine mussel, Mytilus edulis. These membranes contained two distinct pathways for cotransport of Na+ and alpha-neutral amino acids. The major pathway in mussel gill BBMV was the alanine-lysine
Mitsuyoshi Yoshimoto et al.
Cancer science, 109(5), 1617-1626 (2018-03-03)
In boron neutron capture therapy (BNCT), 10 B-4-borono-L-phenylalanine (BPA) is commonly used as a 10 B carrier. PET using 4-borono-2-18 F-fluoro-phenylalanine (18 F-FBPA PET) has been performed to estimate boron concentration and predict the therapeutic effects of BNCT; however, the
1500368
Meira W, et al.
Cancers, 13(6), 1434-1434 (2021)
J R Bading et al.
Nuclear medicine and biology, 23(6), 779-786 (1996-08-01)
The A system of amino acid transport is concentrative and thought to be a regulator of cell growth. The [11C]methyl alpha-aminoisobutyric acid (MeAIB) is prospectively an ideal tracer for transport measurements with PET, as it is not metabolized and concentrates
Nora Sandow et al.
Epilepsia, 50(4), 849-858 (2009-01-30)
Glutamine (GLN) is a precursor for synthesis of glutamate and gamma-aminobutyric acid (GABA) and has been found in the cerebrospinal fluid (CSF) at mean concentrations of 0.6 mM. Experiments on slices are usually performed in artificial CSF (aCSF) kept free

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service