Skip to Content
Merck
All Photos(2)

Key Documents

A4562

Sigma-Aldrich

5-(N,N-Dimethyl)amiloride hydrochloride

Synonym(s):

3-Amino-N-(aminoiminoethyl)-5-(dimethylamino)-6-chloropyrazinecarboxamide hydrochloride, DMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12ClN7O · HCl
CAS Number:
Molecular Weight:
294.14
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

Cl.CN(C)c1nc(N)c(nc1Cl)C(=O)\N=C(\N)N

InChI

1S/C8H12ClN7O.ClH/c1-16(2)6-4(9)13-3(5(10)14-6)7(17)15-8(11)12;/h1-2H3,(H2,10,14)(H4,11,12,15,17);1H

InChI key

IIUPTHVVXMBJMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-(N,N-Dimethyl)amiloride hydrochloride is a selective blocker of Na2/H+ antiport. 5-(N,N-Dimethyl)amiloride hydrochloride has been investigated for its ability to reduce inflammatory pain.

Biochem/physiol Actions

Selective blocker of Na+/H+ antiport; IC50 = 6.9 μM.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D F Zhao et al.
Nature communications, 8, 14067-14067 (2017-02-22)
The terrestrial vegetation emits large amounts of volatile organic compounds (VOC) into the atmosphere, which on oxidation produce secondary organic aerosol (SOA). By acting as cloud condensation nuclei (CCN), SOA influences cloud formation and climate. In a warming climate, changes
Matija Lovrak et al.
Nature communications, 8, 15317-15317 (2017-06-06)
Self-assembly provides access to a variety of molecular materials, yet spatial control over structure formation remains difficult to achieve. Here we show how reaction-diffusion (RD) can be coupled to a molecular self-assembly process to generate macroscopic free-standing objects with control
Karin Engström et al.
Archives of toxicology, 91(5), 2067-2078 (2016-11-14)
Arsenic, a carcinogen with immunotoxic effects, is a common contaminant of drinking water and certain food worldwide. We hypothesized that chronic arsenic exposure alters gene expression, potentially by altering DNA methylation of genes encoding central components of the immune system.
Yered Pita-Juárez et al.
PLoS computational biology, 14(3), e1006042-e1006042 (2018-03-20)
A goal of genomics is to understand the relationships between biological processes. Pathways contribute to functional interplay within biological processes through complex but poorly understood interactions. However, limited functional references for global pathway relationships exist. Pathways from databases such as
Peter A Alpert et al.
Scientific reports, 7(1), 12693-12693 (2017-10-06)
Organic interfaces that exist at the sea surface microlayer or as surfactant coatings on cloud droplets are highly concentrated and chemically distinct from the underlying bulk or overlying gas phase. Therefore, they may be potentially unique locations for chemical or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service