Skip to Content
Merck
All Photos(1)

Key Documents

SML1006

Sigma-Aldrich

EGA

≥98% (HPLC)

Synonym(s):

2-[(4-Bromophenyl)methylene]-N-(2,6-dimethylphenyl)-hydrazinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16BrN3O
CAS Number:
Molecular Weight:
346.22
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/vial, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

EGA is a small molecule that blocks the entry of anthrax lethal toxin by inhibiting trafficking pathways in acidified endosomes, without affecting endosome pH. EGA blocks trafficking of other toxins, including diphtheria toxin, but does not inhibit trafficking of ricin. The compound EGA attenuates lysosomal targeting and degradation of the EGF receptor, but does not block does not block endosomal recycling of transferrin.
EGA is an inhiitor of endosomal trafficking.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel J Dobson et al.
Antiviral research, 179, 104819-104819 (2020-05-12)
During virus entry, members of the Polyomaviridae transit the endolysosomal network en route to the endoplasmic reticulum (ER), from which degraded capsids escape into the cytoplasm and enter the nucleus. Emerging evidence suggests that viruses require both endosomal acidification and
Yu Wu et al.
The FEBS journal, 287(15), 3184-3199 (2020-01-05)
The endo-lysosome system is involved in endocytosis, protein sorting, and degradation as well as autophagy. Numerous toxins and pathogens exploit this system to enter host cells and exert their deleterious effects. Modulation of host endo-lysosome pathway may restrict multiple toxins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service