Skip to Content
Merck
All Photos(1)

Key Documents

A3437

Sigma-Aldrich

Anti-Human IgM (μ-chain specific)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Goat Anti-Human IgM (μ-chain specific)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

direct ELISA: 1:30,000
dot blot: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Human IgMs are the initial immunoglobulin isotypes that appear in blood in response to the first exposure to external antigens. These immunoglobulins have been implicated in CNS myelin repair. Increased levels of secretory IgM have been linked to rhino-conjunctivitis . Anti-Human IgM (μ-chain specific)-Alkaline Phosphatase antibody is specific for human IgM when tested against human IgA, IgG, IgM, Bence Jones κ, and λ myeloma proteins.
Immunoglobulin M (IgM) antibodies appear early in the course of infections. IgM antibodies are responsible for agglutination of red blood cells in mis-matched blood transfusions. The level of IgM may vary with the status of disease or infection.
Alkaline Phosphatase is an enzyme that catalyzes the conversion of chromogenic substrates such as p-nitrophenylphosphate (PNPP); chemiluminescent substrates such as CDP-Star® and fluorogenic substrates such as 4-methylumbelliferyl phosphate (4-MUP) into detectable chromophores, light-emitters or fluorescers, respectively.

Immunogen

Human IgM

Application

Anti-Human IgM (μ-chain specific)-Alkaline Phosphatase antibody is suitable for use in ELISA.
Goat Anti-Human IgM (μ-chain specific)-Alkaline Phosphatase antibody can be used for dot blot and western blot applications at 1:30,000.
Goat polyclonal anti-Human IgM (μ-chain specific)−Alkaline Phosphatase antibody may be used to detect and quantitate the level of IgM in human serum and biological fluids via chromogenic, chemoluminescent or fluorogenic immunochemical or immunohistochemical techniques.
IgM is a glycoprotein with 5 n-linked glycosylation sites on the heavy chain. An ELISA assay was performed to identify glycosylated forms of IgM that bind to lectin. Alkaline phosphatase conjugated goat anti-human IgM was used as the secondary at 1:2000 and developed using p-nitrophenyl substrate (Sigma).
IgM is a glycoprotein with 5 n-linked glycosylation sites on the heavy chain. An ELISA assay was performed to identify glycosylated forms of IgM that bind to lectin. Alkaline phosphatase conjugated goat anti-human IgM was used as the secondary at 1:2000 and developed using p-nitrophenyl substrate (Sigma). Arnold, J. (2005) Human serum IgM glycosylation: identification of glycoforms that can bind to mannan-binding lectin. JBC, 280: 29080-29087.

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 10 mM glycine, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide

Legal Information

CDP-Star is a registered trademark of Tropix, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Petry
Infection, 26(1), 7-10 (1998-03-20)
In a seroprevalence study including 495 sera from persons of all age-groups, the presence of anti-Cryptosporidium parvum antibodies was evaluated in an enzyme immunoassay. Despite the fact that C. parvum is only found in approximately 2% of patients with diarrhea
Helena Cerutti et al.
Journal of clinical laboratory analysis, 35(4), e23735-e23735 (2021-02-21)
The rapid spread of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has generated a pandemic with alarming rates of fatality worldwide. This situation has had a major impact on clinical laboratories that have attempted to answer the urgent need for
Michael Sciaudone et al.
IJID regions, 7, 164-169 (2023-04-11)
Racial and ethnic minorities have borne a disproportionate burden from coronavirus disease 2019 (COVID-19). Certain essential occupations, including food processing and farm work, employ large numbers of Hispanic migrant workers and have been shown to carry an especially high risk
S Eichinger et al.
Journal of thrombosis and haemostasis : JTH, 16(1), 31-35 (2017-10-19)
Essentials Natural antibodies to oxidation-specific epitopes have antithrombotic properties. We evaluated the relation between natural IgM and IgG antibodies and the venous thrombosis risk. Risk of recurrent thrombosis was higher in patients with low natural IgM antibody levels. The protective
Miryan Margot Sánchez-Jiménez et al.
Heliyon, 6(7), e04393-e04393 (2020-07-21)
Brucella canis, a Gram-negative coccobacilli belonging to the genus Brucellae, is a pathogenic bacterium that can produce infections in dogs and humans. Multiple studies have been carried out to develop diagnostic techniques to detect all zoonotic Brucellae. Diagnosis of Brucella

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service