Skip to Content
Merck
All Photos(1)

Documents

B6149

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl phosphate disodium salt

chromogenic, ≥98% (HPLC), powder

Synonym(s):

5-BCIP disodium salt, BCIP®, X-phosphate disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H4BrClNO4P · 2Na
CAS Number:
Molecular Weight:
370.43
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

5-Bromo-4-chloro-3-indolyl phosphate disodium salt, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

solubility

H2O: 20 mg/mL
DMF: insoluble

storage temp.

−20°C

SMILES string

[Na+].[Na+].[O-]P([O-])(=O)Oc1c[nH]c2ccc(Br)c(Cl)c12

InChI

1S/C8H6BrClNO4P.2Na/c9-4-1-2-5-7(8(4)10)6(3-11-5)15-16(12,13)14;;/h1-3,11H,(H2,12,13,14);;/q;2*+1/p-2

InChI key

OAZUOCJOEUNDEK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

5-Bromo-4-chloro-3-indolyl phosphate disodium salt has been used to detect protein-antibody complexes. It has also been used for tissue staining in alkaline phosphatase histochemistry.
For the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.

BCIP is provided in two salt forms: the disodium salt which is soluble in water and the p-toluidine form which is soluble in dimethylformamide. These salt forms may be used to prepare a stock solution that in combination with NBT and a reaction buffer, form a substrate solution for alkaline phosphatase. This substrate system, when incubated with alkaline phosphatase, produces an insoluble NBT diformazan product that is easily observable with its purple color. For added convenience, a mixture of BCIP and NBT is provided in an easily dissolvable tablet form or as a ready-to-use liquid.

Substrates

Histochemical substrate for alkaline phosphatase.

Legal Information

BCIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manli Qi et al.
Journal of bacteriology, 193(10), 2498-2509 (2011-03-29)
The Chlamydia-specific hypothetical protein CT795 was dominantly recognized by human antisera produced during C. trachomatis infection but not by animal antisera raised against dead chlamydia organisms. The immundominant region recognized by the human antibodies was mapped to the N-terminal fragment
P E Høyer et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 44(8), 855-863 (1996-08-01)
We describe a quantitative histochemical method for demonstration of five N-acetyl-glucosamine binding lectins in the syncytiotrophoblast of human term placenta. The method employs biotinylated lectins and alkaline phosphatase-conjugated avidin. The alkaline phosphatase activity is detected by using 5-bromo-4-chloro-indoxyl phosphate as
Shailesh Kumar et al.
PLoS genetics, 15(6), e1008219-e1008219 (2019-06-27)
Fes1 is a conserved armadillo repeat-containing Hsp70 nucleotide exchange factor important for growth at high temperature, proteasomal protein degradation and prion propagation. Depleting or mutating Fes1 induces a stress response and causes defects in these processes that are ascribed solely
Thermal preference ranges correlate with stable signals of universal stress markers in Lake Baikal endemic and Holarctic amphipods
Axenov-Gribanov D, et al
PLoS ONE, 11(10), e0164226-e0164226 (2016)
The endosomal protein endotubin is required for enterocyte differentiation
Cox CM, et al.
Cellular and molecular gastroenterology and hepatology, 5(2), 145-156 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service