Skip to Content
Merck
All Photos(1)

Key Documents

A9794

Sigma-Aldrich

Anti-Human IgM (μ-chain specific)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Human IgM (μ-chain specific)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:50,000
dot blot: 1:60,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200
western blot (chemiluminescent): 1:60,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

There are five immunoglobulin classes in humans. Out of these, immunoglobulin M (IgM ) is a high molecular weight protein that has five or six subunits. IgM monomers are made of two heavy chains and two light chains connected by a disulfide bond. Human serum shows low concentration of IgM monomers. It has a high carbohydrate content of about 12%. It is the first immunoglobulin produced by neonates.

Specificity

Goat polyclonal anti-Human IgM (μ-chain specific)-Alkaline Phosphatase antibody is specific for human IgM when tested against human IgA, IgG, IgM, Bence Jones kappa, and lambda myeloma proteins. The conjugate shows no reactivity with mouse or rat IgG.

Application

IgM is a glycoprotein with 5 n-linked glycosylation sites on the heavy chain. An ELISA assay was performed to identify glycosylated forms of IgM that bind to lectin. Alkaline phosphatase conjugated goat anti-human IgM was used as the secondary at 1:2000 and developed using p-nitrophenyl substrate (Sigma).

Biochem/physiol Actions

Immunoglobulin M (IgM) acts as an antigen specific part of the B cell antigen receptor on the surface of B lymphocytes that are not stimulated, in its monomeric form. Polymeric IgM molecules also serve as important activators of the classical complement cascade. IgM is essential in agglutination and cytolytic reactions.
Immunoglobulin M (IgM) antibodies appear early in the course of infections. IgM antibodies are responsible for agglutination of red blood cells in mis-matched blood transfusions. The level of IgM may vary with the status of disease or infection. Alkaline Phosphatase is an enzyme that catalyzes the conversion of chromogenic substrates such as p-nitrophenylphosphate (PNPP); chemiluminescent substrates such as CDP-Star® and fluorogenic substrates such as 4-methylumbelliferyl phosphate (4-MUP) into detectable chromophores, light-emitters or fluorescers, respectively.

Other Notes

No cross-reaction with mouse and rat IgG

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide

Legal Information

CDP-Star is a registered trademark of Tropix, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A V Sokoloff et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 3(6), 821-830 (2001-06-16)
Our previous study indicated that normal serum contains complement-fixing natural IgM antibodies reacting with a large variety of randomly generated protein carboxy-termini. Here we show that the "carboxy-terminal" IgM (C-IgM) antibodies specifically react with short peptide sequences located immediately at
M Sämpi et al.
European journal of clinical investigation, 39(6), 517-526 (2009-06-06)
Involvement of low density lipoprotein (LDL) immune complexes (ICs) in atherogenesis has been proposed. Human FcgammaRIIa receptor (CD32) plays a crucial role in the phagocytosis of IgG(2) ICs and a functional point mutation 131His/Arg diminishes IgG(2) binding to the receptor.
Emily R Adams et al.
Wellcome open research, 5, 139-139 (2021-03-23)
Background: The COVID-19 pandemic caused >1 million infections during January-March 2020. There is an urgent need for reliable antibody detection approaches to support diagnosis, vaccine development, safe release of individuals from quarantine, and population lock-down exit strategies. We set out
Krista G Haanstra et al.
PloS one, 8(8), e71549-e71549 (2013-08-27)
The overlapping epidemiology of multiple sclerosis (MS) and Epstein-Barr virus (EBV), the increased risk to develop MS after infectious mononucleosis (IM) and the localization of EBV-infected B-cells within the MS brain suggest a causal link between EBV and MS. However
Krista G Haanstra et al.
Journal of immunology (Baltimore, Md. : 1950), 194(4), 1454-1466 (2015-01-16)
Costimulatory and coinhibitory receptor-ligand pairs on T cells and APC control the immune response. We have investigated whether selective blockade of CD28-CD80/86 costimulatory interactions, which preserves the coinhibitory CTLA4-CD80/86 interactions and the function of regulatory T (Treg) cells, abrogates the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service