Skip to Content
Merck
All Photos(1)

Documents

A0420

Sigma-Aldrich

Anti-Human IgM (μ-chain specific)−Peroxidase antibody produced in goat

affinity isolated antibody

Synonym(s):

Goat Anti-Human IgM (μ-chain specific)−HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

technique(s)

direct ELISA: 1:50,000
dot blot: 1:100,000 (chemiluminescent)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

B cells predominantly secrete IgM antibodies as pentamers, but may also secrete them as hexamers which are capable of activating the complement cascade more efficiently.
Goat anti-human IgM (mu-chain specific)-peroxidase antibody is specific for human IgM and does not bind to other human Igs. Furthermore, the antibody does not react with mouse and rat IgG. Hence, the antibody conjugate yields reduced background with mouse or rat samples.

Immunogen

human IgM

Application

Anti- Human IgM was used in an ELISA assay to determine the concentration of IgM secreted by CH12 B lymphoma cells stimulated by LPS.
Goat anti-human IgM (mu-chain specific)-peroxidase antibody can be used:
  • for dot blot (1:100,000) and immunohistochemistry (1:200 using formalin-fixed, paraffin-embedded sections) applications.
  • in immunoblotting
  • sandwich immunoassay

Biochem/physiol Actions

IgM plays a key role in engulfing of the apoptotic cells. A reduction in serum IgM levels leads to increased autoimmune response and higher risk for infections. IgM displays polyreactive and autoreactive functionality. It plays a key role in tissue homeostasis by mediating clearance of tissue based molecules.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 0.05% MIT

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cancer marker detection in human serum with a point-of-care low-cost system
Grosso P, et al.
Sensors and Actuators B, Chemical, 147(2), 475-480 (2010)
Fully human IgG and IgM antibodies directed against the carcinoembryonic antigen (CEA) Gold 4 epitope and designed for radioimmunotherapy (RIT) of colorectal cancers
Garambois V, et al.
BMC Cancer, 4(1), 75-75 (2004)
Kioa L Wijnsma et al.
Pediatric nephrology (Berlin, Germany), 31(11), 2163-2170 (2016-06-01)
In the majority of pediatric patients, the hemolytic-uremic syndrome (HUS) is caused by an infection with Shiga toxin-producing Escherichia coli (STEC), mostly serotype O157. It is important to discriminate between HUS caused by STEC and complement-mediated HUS (atypical HUS) due
Asfia Qureshi et al.
Mycopathologia, 173(5-6), 419-425 (2011-10-06)
We have developed an ELISA to determine the value of anti-glucosylceramide antibody for the prediction of disseminated cryptococcosis in immunocompromised subjects and performed a clinical prospective study at the Medical University of South Carolina. The study enrolled a total of
María Luz Galván-Ramírez et al.
Transplant infectious disease : an official journal of the Transplantation Society, 21(5), e13139-e13139 (2019-07-05)
Solid organ transplant recipients are highly susceptible to Toxoplasma gondii infection. We aimed to describe the 12-month follow-up risk of seroconversion in renal transplant recipients. Anti-T gondii antibodies were investigated in donors and recipients of renal transplants. In donors, anti-T gondii were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service