Skip to Content
Merck
All Photos(1)

Key Documents

A3316

Sigma-Aldrich

Anti-Human IgG (Fc Specific)−Agarose antibody produced in goat

affinity isolated antibody, PBS suspension

Synonym(s):

Goat anti-human IgG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

agarose conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

PBS suspension

species reactivity

human

should not react with

rat, mouse

technique(s)

Ouchterlony double diffusion: suitable

capacity

2 mg/mL, resin binding capacity (human IgG)

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders.
Goat Anti-Human IgG (Fc Specific)-Agarose antibody is specific for the Fc fragment of human IgG when tested against purified human IgA, IgG, IgM, Fc, and kappa and lambda light chains. No reactivity with mouse or rat IgG is seen by Ouchterlony Double Diffusion (ODD), prior to agarose bead coupling.

Immunogen

Fc fragment of human IgG.

Application

Anti-Human IgG (Fc Specific)-Agarose antibody produced in goat has also been used to purify galanin 1 (Gala1), 3Gal-substituted P-selectin glycoprotein ligand-1 (PSGL-1)/hIgG1 from supernatants of transfected COS cells.
Goat Anti-Human IgG (Fc Specific)-Agarose antibody is suitable for use in Ouchterlony double diffusion. The antibody has also been used for affinity purification assays.
Immunoprecipitation assays were performed using goat anti-human IgG (Fc specific) crosslinked to agarose beads. The beads were diluted in 1 ml of 5% FBS/DMEM for 2 hours at 4 degrees.

Other Notes

Antibody adsorbed with mouse and rat IgG

Physical form

Suspension in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Xu et al.
Immunity, 13(1), 37-45 (2000-08-10)
All rearranging antigen receptor genes have one or two highly diverse complementarity determining regions (CDRs) among the six that typically form the ligand binding surface. We report here that, in the case of antibodies, diversity at one of these regions
Rebecca J Brown et al.
The Journal of clinical endocrinology and metabolism, 102(6), 1789-1791 (2016-12-03)
Hyperinsulinemia can lead to pathologic ovarian growth and androgen production. A 29-year-old woman developed an autoantibody to the insulin receptor (type B insulin resistance), causing extreme insulin resistance and hyperinsulinemia. Testosterone levels were elevated to the adult male range. Treatment
Guang Song et al.
Molecular & cellular proteomics : MCP, 17(2), 349-356 (2017-11-17)
Zika virus (ZIKV) and dengue virus (DENV) are closely related flaviviruses that cause widespread, acute febrile illnesses, notably microcephaly for fetuses of infected pregnant women. Detecting the viral cause of these illnesses is paramount to determine risks to patients, counsel
Manuel Albanese et al.
Cell reports. Medicine, 5(4), 101483-101483 (2024-04-06)
Immune cell phenotyping frequently detects lineage-unrelated receptors. Here, we report that surface receptors can be transferred from primary macrophages to CD4 T cells and identify the Fcγ receptor CD32 as driver and cargo of this trogocytotic transfer. Filamentous CD32+ nanoprotrusions deposit
M Kristen Demoruelle et al.
Arthritis research & therapy, 23(1), 163-163 (2021-06-08)
Mucosal sites are hypothesized to play a role in the development of rheumatoid arthritis (RA). Since serum anti-peptidylarginine deiminase (PAD)4 antibodies, including a subset that cross-react with PAD3 (PAD3/4), are specific for RA and associate with severe disease, we sought

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service