Skip to Content
Merck
All Photos(1)

Key Documents

A9935

Sigma-Aldrich

Anti-Human IgM (μ-chain specific)−Agarose antibody produced in goat

affinity isolated antibody, PBS suspension

Synonym(s):

Anti Human IgM Antibody - Anti-Human IgM (μ-chain specific)-Agarose antibody produced in goat, Anti Human Igm Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

agarose conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

PBS suspension

technique(s)

Ouchterlony double diffusion: suitable
immunoelectrophoresis: suitable

capacity

2-3 mg/mL, resin binding capacity (human IgM)

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Goat polyclonal anti-Human IgM (μ-chain specific)−Agarose antibody specific for human IgM when tested against purified human IgA, IgG, IgM, and kappa and lambda light chains. No reactivity with mouse or rat IgG is observed by Ouchterlony Double Diffusion (ODD), prior to agarose bead coupling.
IgM is a highly conserved antibody in vertebrates and is expressed early during immune response. It exists as a pentamer and is secreted by peritoneal B cells.
Immunoglobulin M (IgM) antibodies appear early in the course of infections. IgM antibodies are responsible for agglutination of red blood cells in mis-matched blood transfusions. The level of IgM may vary with the status of disease or infection.
Cyanogen bromide activated cross-linked beaded agarose is frequently conjugated to proteins such as immunoglobulins to facilitate their use in physicochemical processes such as precipitation, absorption and clarification.

Application

Anti-Human IgM (μ-chain specific)−Agarose antibody produced in goat has been used in affinity chromatography purification of human IgM.
Goat polyclonal anti-Human IgM (μ-chain specific)−Agarose antibody conjugate is used as an immuno-adsorbent to affinity purify antibodies, to remove species specific cross-reacting antibodies or to remove contaminating antibodies from an antiserum preparation. It has been used for IgM depletion from normal human serum from fasted patients.

Biochem/physiol Actions

IgM plays a key role in the engulfing of apoptotic cells. A reduction in serum IgM levels leads to increased autoimmune response and higher risk for infections. IgM displays polyreactive and autoreactive functionality. It plays a key role in tissue homeostasis by mediating clearance of tissue based molecules. IgM has sites for N-linked glycosylation, with sugars namely, mannose, galactose, N-acetyl glucosamine and sialic acid. IgM linked agarose resins have been tested for efficient conjugate binding.

Other Notes

Antibody adsorbed with mouse and rat IgG

Physical form

Suspension in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rationally designed mimotope library for profiling of the human IgM repertoire
Pashov AD, et al.
bioRxiv, 308973-308973 (2018)
Jan Koopmans et al.
Cell transplantation, 13(6), 631-637 (2005-01-15)
Xenografting pig fetal ventral mesencephalic (pfVM) cells to repair the dopamine deficit in patients with Parkinson's disease is the focus of both experimental and clinical investigations. Although there have been marked advances in the experimental and even clinical application of
Anastas Pashov et al.
Frontiers in immunology, 10, 2796-2796 (2019-12-19)
Specific antibody reactivities are routinely used as biomarkers, but the antibody repertoire reactivity (igome) profiles are still neglected. Here, we propose rationally designed peptide arrays as efficient probes for these system level biomarkers. Most IgM antibodies are characterized by few
S Kløverpris et al.
Molecular human reproduction, 19(11), 756-763 (2013-06-28)
Pregnancy-associated plasma protein-A (PAPP-A) and PAPP-A2, two homologous metzincin metalloproteases, are both tightly linked to regulation within the insulin-like growth factor (IGF) system because of their specific cleavage of IGF binding proteins. Recent studies suggest that PAPP-A may be involved
Whitney Bullard et al.
Virulence, 3(1), 72-76 (2012-01-31)
The haptoglobin-hemoglobin receptor (HpHbR) of African trypanosomes plays a critical role in human innate immunity against these parasites. Localized to the flagellar pocket of the veterinary pathogen Trypanosoma brucei brucei this receptor binds Trypanosome Lytic Factor-1 (TLF-1), a subclass of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service