Skip to Content
Merck
All Photos(5)

Key Documents

H9658

Sigma-Aldrich

Monoclonal Anti-HA antibody produced in mouse

clone HA-7, ascites fluid

Synonym(s):

Monoclonal Anti-HA, HA Antibody - Monoclonal Anti-HA antibody produced in mouse, Ha Antibody Sigma, Sigma Ha Antibody, Anti-HA, Anti-Influenza Hemagglutinin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

HA-7, monoclonal

contains

15 mM sodium azide

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:20,000 using extracts from 293T (human embryonic kidney) cells expressing a recombinant HA-fusion protein

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

The gene encoding influenza virus hemagglutinin (HA) precursor is mapped on segment 4. It encodes a 76kDa surface glycoprotein which exists as trimers on the viral envelope.

Specificity

The antibody recognizes native as well as denatured-reduced forms of HA-tagged proteins and is reactive with N- or C-terminal HA-tagged fusion proteins expressed in E. coli or in mammalian cells.

Immunogen

synthetic peptide corresponding to a fragment of human influenza virus hemagglutinin (HA) known as HA-tag, conjugated to KLH

Application

Monoclonal Anti-HA antibody produced in mouse has been used in 293T cells for:
  • immunoprecipitation in human embryonic kidney 293 cells
  • immunoblotting in HeLa cells
  • immunopurification in human embryonic kidney 293 cells
  • immunostaining for electron microscopy in mouse embryonic fibroblasts
  • in immunocytochemistry of hippocampal slices
Monoclonal Anti-HA antibody produced in mouse has been used in:
  • Immunoblotting, to study the expression of fusion protein
  • Coimmunoprecipitation assay
  • Immunoblotting of lysate protein

Biochem/physiol Actions

Hemagglutinin (HA) is the surface glycoprotein of influenza virus. HA is crucial for virus entry and interaction with N-acetylneuraminic acid (Neu5Ac) in host cell surface. HA coordinates with neuraminidase (NA) and facilitates viral entry into host. HA exists as a trimer and each monomer has stalk transmembrane and globular regions. The globular region contains carbohydrate side chains crucial for infecting host.
Influenza virus hemagglutinin (HA) acts as a key immunogen in avian influenza virus (AIV) vaccines. Activated HA facilitates virus attachment to oligosaccharide receptors on host cell surface and is involved in penetration through fusion following conformational modifications in the endolysosomal compartment. 226 and 228 residues in hemagglutinin (HA) of the H3 subtype play a key role in host range restriction and receptor specificity. Influenza virus hemagglutinin (HA) is responsible for 1918 influenza pandemic.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.
For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Solutions at working dilution should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Visualization of Alternative Functional Configurations of Influenza Virus Hemagglutinin Facilitates Rapid Selection of Complementing Vaccines in Emergency Situations.
Metwally A and Yousif A
International Journal of Molecular Sciences, 18(4), 766-766 (2017)
Sumoylation of histone deacetylase 1 regulates MyoD signaling during myogenesis
Joung H, et al.
Experimental & Molecular Medicine, 50(1), e427-e427 (2018)
A Hox-Eya-Pax complex regulates early kidney developmental gene expression.
Gong K Q, et al.
Molecular and Cellular Biology, 27(21), 7661-7668 (2007)
Synaptic targeting and functional modulation of GluK1 kainate receptors by the auxiliary neuropilin and tolloid-like (NETO) proteins.
Copits B A, et al.
The Journal of Neuroscience, 31(20), 7334-7340 (2011)
Calmodulin shuttling mediates cytonuclear signaling to trigger experience-dependent transcription and memory
Cohen SM, et al.
Nature Communications, 9(1), 2451-2451 (2018)

Questions

1–4 of 4 Questions  
  1. I want to do an IP using this HA antibody. I know it is produced in mouse but what kind of IgG is it?

    1 answer
    1. This antibody is an IgG1 isotype. Please see the link below to review additional information available in the product datasheet:_x000D_
      https://www.sigmaaldrich.com/US/en/product/sigma/h9658

      Helpful?

  2. What is the isotype and light chain for Monoclonal Anti-HA antibody clone HA-7 produced in mouse?

    1 answer
    1. Monoclonal Anti-HA antibody clone HA-7 is mouse IgG1, kappa light chain.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. What is the recommended dilution of Product H9658 - HA antibody for immunoprecipitation and for indirect immunostaining?

    1 answer
    1. H9658 - Monoclonal Anti-HA antibody produced in mouse is the ascites fluid. It is not purified. We only test this product in western blotting.  This product contains 15-55 mg/ml total protein with 4-7 mg/ml specific antibody. H3663 - Monoclonal Anti-HA antibody produced in mouse as the purified immunoglobulin. We test this product for immunoprecipitation using 0.5-2.0 μg and for immunocytochemistry using 1.0-2.0 μg/mL.These ranges would be a good starting concentration. In order to obtain best results in different techniques and preparations, we recommend determining optimal working dilution by titration test.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service