Skip to Content
Merck
All Photos(4)

Key Documents

A2545

Sigma-Aldrich

Adenine hemisulfate salt

suitable for plant cell culture, BioReagent, ≥99%

Synonym(s):

6-Aminopurine hemisulfate salt, Adenine sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H5N5 · 1/2H2SO4
CAS Number:
Molecular Weight:
184.17
Beilstein:
3820263
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Quality Level

Assay

≥99%

form

powder

technique(s)

cell culture | plant: suitable

solubility

0.5 M HCl: 10 mg/mL

application(s)

agriculture

SMILES string

OS(O)(=O)=O.Nc1ncnc2[nH]cnc12.Nc3ncnc4[nH]cnc34

InChI

1S/2C5H5N5.H2O4S/c2*6-4-3-5(9-1-7-3)10-2-8-4;1-5(2,3)4/h2*1-2H,(H3,6,7,8,9,10);(H2,1,2,3,4)

InChI key

LQXHSCOPYJCOMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adenine hemisulfate salt is used as a component of minimal medium and yeast extract dextrose, peptone and adenine (YPAD) medium component for culturing yeast.

Application

Adenine hemisulfate salt has been used as a component of medium D and medium SP for protoplast culture
Adenine may be used as a plant cell culture additive. Adenine Sulphate (25mg/L) in the culture media such as Murashige & Skoog media improves the frequency of multiple shoot production and recovered the chlorotic symptoms of the leaves.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Bulter et al.
Applied and environmental microbiology, 69(2), 987-995 (2003-02-07)
Laccase from Myceliophthora thermophila (MtL) was expressed in functional form in Saccharomyces cerevisiae. Directed evolution improved expression eightfold to the highest yet reported for a laccase in yeast (18 mg/liter). Together with a 22-fold increase in k(cat), the total activity
Ryan R Cochrane et al.
Biology, 9(11) (2020-10-30)
Algae are attractive organisms for biotechnology applications such as the production of biofuels, medicines, and other high-value compounds due to their genetic diversity, varied physical characteristics, and metabolic processes. As new species are being domesticated, rapid nuclear and organelle genome
Sanghamitra Samantaray et al.
Revista de biologia tropical, 59(1), 435-445 (2011-04-26)
Chlorophytum arundinaceum is an important medicinal plant and its tuberous roots are used for various health ailment treatments. It has become an endangered species in the Eastern Ghats, and a rare medicinal herb in India, due to its excessive collection
Protoplast technology in genome manipulation of potato through somatic cell fusion
Somatic Genome Manipulation, 217-235 (2015)
Santosh Kumar Rai et al.
Applied biochemistry and biotechnology, 162(7), 1929-1937 (2010-05-18)
An efficient in vitro process for rapid production of cloned plants of Uraria picta has been developed employing nodal stem segments taken from field-grown plants. Explants showed bud-break followed by regeneration of shoots with restricted growth within 12 days on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service