Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adenine hemisulfate salt
Synonym(s):
6-Aminopurine hemisulfate salt, Adenine sulfate salt
Linear Formula:
C
5
H
5
N
5
· 1/2H
2
SO
4
CAS Number:
321-30-2
Molecular Weight:
184.17
EC Number:
206-286-5
Products
Brand
Sigma-Aldrich (3)
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A9126
Adenine hemisulfate salt,
≥99%
Expand
View Pricing
A3159
Adenine hemisulfate salt,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
A2545
Adenine hemisulfate salt,
suitable for plant cell culture, BioReagent, ≥99%
Expand
View Pricing