Skip to Content
Merck
All Photos(1)

Key Documents

91211

Sigma-Aldrich

Oxyresveratrol

≥97.0% (HPLC)

Synonym(s):

(E)-2,3′,4,5′-Stilbenetetrol, 2,3′,4,5′-Tetrahydroxy-trans-stilbene, 4-[(1E)-2-(3,5-Dihydroxyphenyl)ethenyl]-1,3-benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O4
CAS Number:
Molecular Weight:
244.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥97.0% (HPLC)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

Oc1ccc(\C=C\c2cc(O)cc(O)c2)c(O)c1

InChI

1S/C14H12O4/c15-11-4-3-10(14(18)8-11)2-1-9-5-12(16)7-13(17)6-9/h1-8,15-18H/b2-1+

InChI key

PDHAOJSHSJQANO-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxyresveratrol (OXY or ORV) is a naturally occurring polyphenolic stilbene that is predominantly present in the heartwood of many plants. It is primarily extracted from Artocarpus lakoocha Roxb. plant, and is one of the main constituents of this plant. ORV has a very simple chemical structure with a trans-1,2-diphenylethylene in its core and two hydroxyls (OH) groups attached to each aromatic ring.

Application

Oxyresveratrol has been used:
  • with poly(lactic-co-glycolic acid) (PLGA) nanoparticles to test its antioxidant effects and to investigate its role in the inhibition of superoxide anion (O2-) generation
  • to test its effects as an anti-neuroinflammatory agent on interleukin-1 beta (IL-1β)-induced HMC3 human microglial cell line
  • as a reference standard to determine its existence in Artocarpus lakoocha Roxb. (AL) extract using high-performance liquid chromatography (HPLC)

Biochem/physiol Actions

Oxyresveratrol is well-known due to its diverse biological and pharmacological properties. It is a potent antioxidant and anti-inflammatory agent. ORV also exerts neuroprotective activities and protects against cerebral ischemia, stroke, and several neurodegenerative disorders. It also displays anti-cancer, antimicrobial, hepatoprotective properties and protects against several other metabolic disorders. It is also a potent tyrosinase inhibitor.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Hee Choi et al.
International journal of molecular sciences, 20(1) (2018-12-24)
The phytochemical oxyresveratrol has been shown to exert diverse biological activities including prevention of obesity. However, the exact reason underlying the anti-obese effects of oxyresveratrol is not fully understood. Here, we investigated the effects and mechanism of oxyresveratrol in adipocytes
Kittisak Likhitwitayawuid
Molecules (Basel, Switzerland), 26(14) (2021-07-25)
Oxyresveratrol has recently attracted much research attention due to its simple chemical structure and diverse therapeutic potentials. Previous reviews describe the chemistry and biological activities of this phytoalexin, but additional coverage and greater accessibility are still needed. The current review
Shaida A Andrabi et al.
Brain research, 1017(1-2), 98-107 (2004-07-21)
Oxidative stress is one of the major pathological factors in the cascade that leads to cell death in cerebral ischemia. Here, we investigated the neuroprotective effect of a naturally occurring antioxidant, oxyresveratrol, to reduce brain injury after cerebral stroke. We
Yi-Ching Chen et al.
BMC complementary and alternative medicine, 13, 45-45 (2013-02-26)
Morus alba has long been used in traditional Chinese medicine to treat inflammatory diseases; however, the scientific basis for such usage and the mechanism of action are not well understood. This study investigated the action of M. alba on leukocyte
Mounir Maafi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 188, 64-71 (2017-07-12)
A new Φ-order kinetic method was proposed in this study for the investigation of trans-cis photoisomerization reaction of Oxyresveratrol (ORVT) subjected to non-isosbestic irradiation. In ethanolic media, it has been proven that forward (Φ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service