M4008
Morin hydrate
powder
Synonym(s):
2′,3,4′,5,7-Pentahydroxyflavone
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C15H10O7 · xH2O
CAS Number:
Molecular Weight:
302.24 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
form
powder
Quality Level
color
yellow
solubility
methanol: 50 mg/mL
SMILES string
O.Oc1ccc(c(O)c1)C2=C(O)C(=O)c3c(O)cc(O)cc3O2
InChI
1S/C15H10O7.H2O/c16-6-1-2-8(9(18)3-6)15-14(21)13(20)12-10(19)4-7(17)5-11(12)22-15;/h1-5,16-19,21H;1H2
InChI key
MYUBTSPIIFYCIU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Morin hydrate has been used:
- to investigate its antitumor functionality using cell viability and apoptosis assay in human cancer cell lines.
- as a fluorescence stain to detect aluminum in motor neurons and lumbar cord.
- to study its protective effect on neuronal hearing loss and on neural stem cells (NSCs) proliferation and differentiation
Biochem/physiol Actions
Morin hydrate is a flavonoid with antioxidant properties. It has been shown to protect cells against oxygen radical damage. Morin not only scavenges oxyradicals, but also moderately inhibits xanthine oxidase, a free-radical generating enzyme. At concentrations of 75-100 micromolar, morin inhibits oxidation of low density lipoprotein (LDL) by free radicals or Cu2+. Morin has been reported to inhibit rat brain phosphatidylinositolphosphate kinase activity in vitro and in vivo.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
T W Wu et al.
Life sciences, 57(3), PL51-PL56 (1995-01-01)
Oxidative modification of low density lipoprotein (LDL) has been suggested to be a risk factor for the development of atherosclerosis. Agents which can protect LDL from oxidation may be useful in preventing atherogenesis. Here, we found that morin hydrate, at
L H Zeng et al.
Biochemistry and cell biology = Biochimie et biologie cellulaire, 75(6), 717-720 (1997-01-01)
Oxygen-derived free radicals are known to injure the endothelium of aorta in diverse disorders. In this study we compared the cytoprotective effects of three flavonoids against oxyradical damage to porcine aortic endothelial cells in vitro. Cultured porcine aortic endothelial cells
L H Zeng et al.
Life sciences, 55(18), PL351-PL357 (1994-01-01)
Cultured rat glomerular mesangial cells were damaged when exposed to oxyradicals generated either from xanthine oxidase plus hypoxanthine, or by superoxide radicals formed from menadione. Morin hydrate is an antioxidant extracted from yellow Brazil wood. When morin hydrate was added
Shan Yu et al.
Inflammation, 43(4), 1293-1303 (2020-03-07)
Mastitis is one of the most common diseases that both affects human and animals. Morin is derived from the member of Moraceae family, which has been used in the treatment of many inflammatory diseases. The purpose of this study was
Aluminum hydroxide injections lead to motor deficits and motor neuron degeneration
Shaw CA and Petrik MS
Journal of Inorganic Biochemistry, 103(11), 1555-1562 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service