Skip to Content
Merck
All Photos(1)

Key Documents

SMB00074

Sigma-Aldrich

Viniferin

≥95% (LC/MS-ELSD)

Synonym(s):

(+)-ε-Viniferin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H22O6
CAS Number:
Molecular Weight:
454.47
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@@H]([C@H](c4c1cc(cc4\C=C\c5ccc(cc5)O)O)c3c(cc(cc3)O)O)c2ccc(cc2)O

InChI

1S/C28H22O6/c29-19-7-2-16(3-8-19)1-4-18-13-22(32)15-25-26(18)27(23-12-11-21(31)14-24(23)33)28(34-25)17-5-9-20(30)10-6-17/h1-15,27-33H/b4-1+/t27-,28+/m0/s1

InChI key

KQXXUMAWOUVEHD-BQYFGGCBSA-N

General description

Natural product derived from plant source.

Application

Viniferin has been used to study its effect on melanin production in melanocyte cultures.

Biochem/physiol Actions

Protein Huntingtin (Htt) mutation, reduces the SIRT3 (sirtuin 3) expression. Viniferin is known to offer protection against Htt mutation by reducing oxidative stress and preventing SIRT3 depletion, which is involved in energy metabolism. In vitro analysis showed that viniferin derivative could prevent amyloid β peptide aggregation.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Aude Pflieger et al.
PloS one, 8(11), e81184-e81184 (2013-12-07)
Polynucleotidyl transferases are enzymes involved in several DNA mobility mechanisms in prokaryotes and eukaryotes. Some of them such as retroviral integrases are crucial for pathogenous processes and are therefore good candidates for therapeutic approaches. To identify new therapeutic compounds and
Advances in Chorea Research and Treatment (2013)
Protective effect of varepsilon-viniferin on beta-amyloid peptide aggregation investigated by electrospray ionization mass spectrometry
Richard T, et al.
Bioorganic & Medicinal Chemistry, 19(10) (2011)
trans-(−)-ε-Viniferin increases mitochondrial sirtuin 3 (SIRT3), activates AMP-activated protein kinase (AMPK), and protects cells in models of Huntington Disease
Fu J, et al.
The Journal of Biological Chemistry, 287(29) (2012)
α-Viniferin Improves Facial Hyperpigmentation via Accelerating Feedback Termination of cAMP/PKA-Signaled Phosphorylation Circuit in Facultative Melanogenesis.
Yun C Y, et al.
Theranostics, 8(7), 2031-2031 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service