Skip to Content
Merck
All Photos(1)

Documents

89204

Supelco

Atto 390 NHS ester

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥90.0% (HPLC)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

extent of labeling

≥90.0%

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 377.0-383.0 nm Amax

suitability

suitable for fluorescence

detection method

fluorometric

storage temp.

−20°C

SMILES string

CC1CC(C)(C)N(CCCC(=O)ON2C(=O)CCC2=O)c3cc4OC(=O)C=C(C)c4cc13

InChI

1S/C24H28N2O6/c1-14-10-23(30)31-19-12-18-16(11-17(14)19)15(2)13-24(3,4)25(18)9-5-6-22(29)32-26-20(27)7-8-21(26)28/h10-12,15H,5-9,13H2,1-4H3

InChI key

ABNZEGUYPHJPQI-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Cuifang Kuang et al.
The Review of scientific instruments, 81(5), 053709-053709 (2010-06-03)
Stimulated emission depletion (STED) microscopy is one of the breakthrough technologies that belong to far-field optical microscopy and can achieve nanoscale spatial resolution. We demonstrate a far-field optical nanoscopy based on continuous wave lasers with different wavelengths, i.e., violet and
Qiang Li et al.
The journal of physical chemistry. B, 115(46), 13643-13649 (2011-10-15)
Fluorescence resonance energy transfer (FRET) using biotinylated β-galactosidase (βGAL) as a donor and Alexa Fluor 350 (AF350) labeled avidin as an acceptor has been investigated by means of steady-state fluorescence and time-resolved fluorescence spectroscopy. The donors are readily paired with
Simon Corrie et al.
Molecular bioSystems, 5(3), 262-268 (2009-02-20)
Herein we report a method for the detection of methylated CpG dinucleotides located within CpG islands in genomic DNA using multiplexed bead-based assays and standard flow cytometry instrumentation. Four CpG "clusters" were identified in the TFPI2 and SPARC CpG islands
Philip Howes et al.
Journal of the American Chemical Society, 132(11), 3989-3996 (2010-02-24)
Semiconducting polymer nanospheres (SPNs) have been synthesized and encapsulated in phospholipid micelles by a solvent evaporation technique. Four different conjugated polymers were used, yielding aqueous dispersions of nanoparticles which emit across the visible spectrum. The synthesis was simple and easily
Gerhild Zauner et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(25), 7085-7090 (2007-06-20)
A fluorescence-based system to sense oxygen in solution is described. The method exploits the sensitivity of the endogenous fluorescence of type-3 copper proteins towards the presence of oxygen by translating the near-UV emission of the protein to label fluorescence in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service