Skip to Content
Merck

Skip To

68321

Atto 390 azide

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C28H41N5O6
Molecular Weight:
543.66
MDL number:
UNSPSC Code:
12352125
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥90% (HPLC)

form

solid

manufacturer/tradename

ATTO-TEC GmbH

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 380-384 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CC(CC(C)(C)N1CCCC(NCCOCCOCCOCCN=[N+]=[N-])=O)C2=C1C=C(OC(C=C3C)=O)C3=C2

InChI

1S/C28H41N5O6/c1-20-16-27(35)39-25-18-24-22(17-23(20)25)21(2)19-28(3,4)33(24)9-5-6-26(34)30-7-10-36-12-14-38-15-13-37-11-8-31-32-29/h16-18,21H,5-15,19H2,1-4H3,(H,30,34)

InChI key

FEBSRHTZZIMYNR-UHFFFAOYSA-N

General description

Atto 390 is a fluorescent label with a coumarin structure. The dye is intended for application in the area of life science, e.g. labeling of DNA, RNA or proteins. Characteristic features of the label are high fluorescence quantum yield, large Stokes-shift, good photostability and low molecular weight.
The azide modification is suitable for reactions with alkyne groups (Huisgen reaction - “Click Chemistry“).

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
685177270978362
suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

form

solid

form

solid

form

solid

form

solid

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

assay

≥90% (HPLC)

assay

≥90% (HPLC)

assay

≥90% (HPLC)

assay

≥90% (HPLC), ≥90% (degree of coupling)

product line

BioReagent

product line

-

product line

BioReagent

product line

BioReagent


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters